Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83893463-83893688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R4
Entrez Gene ID
51167
Gene Name
cytochrome b5 reductase 4
Gene Aliases
NCB5OR, cb5/cb5R, dJ676J13.1
UniGene
Hs.5741
Gene Chromosome Location
Chr.6: 83859651-83960427 on Build GRCh38
Transcripts
NM_016230.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6638765 c.132C>T p.F44F Chr.6: 83893526-83893526 on Build GRCh38
3875867 c.177A>C p.E59D Chr.6: 83893571-83893571 on Build GRCh38
6347111 c.330+8G>T p.? Chr.6: 83893630-83893630 on Build GRCh38
6638764 c.234C>T p.F78F Chr.6: 83893526-83893526 on Build GRCh38
5996784 c.312T>C p.G104G Chr.6: 83893604-83893604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751361804 Chr.6: 83893538-83893538 on Build GRCh38
rs548931086 Chr.6: 83893560-83893560 on Build GRCh38
rs772269331 Chr.6: 83893612-83893612 on Build GRCh38
rs756891340 Chr.6: 83893594-83893594 on Build GRCh38
rs996306407 Chr.6: 83893604-83893604 on Build GRCh38
rs935175201 Chr.6: 83893540-83893540 on Build GRCh38
rs267601144 Chr.6: 83893557-83893557 on Build GRCh38
rs755679834 Chr.6: 83893573-83893573 on Build GRCh38
rs9449718 Chr.6: 83893507-83893507 on Build GRCh38
rs28675051 Chr.6: 83893552-83893552 on Build GRCh38
rs746250827 Chr.6: 83893601-83893601 on Build GRCh38
rs767176877 Chr.6: 83893656-83893656 on Build GRCh38
rs549883802 Chr.6: 83893659-83893659 on Build GRCh38
rs777030261 Chr.6: 83893511-83893511 on Build GRCh38
rs531370161 Chr.6: 83893654-83893654 on Build GRCh38
rs752721075 Chr.6: 83893658-83893658 on Build GRCh38
rs767501609 Chr.6: 83893548-83893548 on Build GRCh38
rs781053747 Chr.6: 83893653-83893653 on Build GRCh38
rs776834820 Chr.6: 83893647-83893647 on Build GRCh38
rs754661226 Chr.6: 83893546-83893546 on Build GRCh38
rs1046332625 Chr.6: 83893498-83893498 on Build GRCh38
rs762100421 Chr.6: 83893527-83893527 on Build GRCh38
rs79475622 Chr.6: 83893524-83893524 on Build GRCh38
rs752528199 Chr.6: 83893555-83893555 on Build GRCh38
rs146871676 Chr.6: 83893595-83893595 on Build GRCh38
rs11539439 Chr.6: 83893570-83893570 on Build GRCh38
rs747956718 Chr.6: 83893655-83893655 on Build GRCh38
rs376929600 Chr.6: 83893553-83893553 on Build GRCh38
rs766332057 Chr.6: 83893535-83893535 on Build GRCh38
rs747437261 Chr.6: 83893622-83893622 on Build GRCh38
rs768903093 Chr.6: 83893640-83893640 on Build GRCh38
rs753537673 Chr.6: 83893587-83893587 on Build GRCh38
rs780637785 Chr.6: 83893619-83893619 on Build GRCh38
rs923688459 Chr.6: 83893616-83893616 on Build GRCh38

View Full Product Details