Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232480178-232480443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECEL1
Entrez Gene ID
9427
Gene Name
endothelin converting enzyme like 1
Gene Aliases
DA5D, DINE, ECEX, XCE
UniGene
Hs.26880
Gene Chromosome Location
Chr.2: 232479827-232487859 on Build GRCh38
Transcripts
NM_001290787.1 NM_004826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4937268 c.2235G>T p.L745L Chr.2: 232480246-232480246 on Build GRCh38
3579091 c.2256G>A p.E752E Chr.2: 232480225-232480225 on Build GRCh38
3991141 c.2266C>T p.R756W Chr.2: 232480215-232480215 on Build GRCh38
6883168 c.2203G>A p.D735N Chr.2: 232480424-232480424 on Build GRCh38
3579090 c.2279G>T p.C760F Chr.2: 232480202-232480202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888224196 Chr.2: 232480341-232480341 on Build GRCh38
rs201486617 Chr.2: 232480240-232480240 on Build GRCh38
rs906039426 Chr.2: 232480310-232480310 on Build GRCh38
rs375638841 Chr.2: 232480383-232480383 on Build GRCh38
rs1029602118 Chr.2: 232480314-232480314 on Build GRCh38
rs200522532 Chr.2: 232480395-232480395 on Build GRCh38
rs776369901 Chr.2: 232480416-232480416 on Build GRCh38
rs762841101 Chr.2: 232480291-232480291 on Build GRCh38
rs755546260 Chr.2: 232480266-232480266 on Build GRCh38
rs1056135287 Chr.2: 232480207-232480207 on Build GRCh38
rs759591149 Chr.2: 232480198-232480198 on Build GRCh38
rs368488157 Chr.2: 232480259-232480259 on Build GRCh38
rs978665800 Chr.2: 232480413-232480413 on Build GRCh38
rs147861587 Chr.2: 232480215-232480215 on Build GRCh38
rs937734530 Chr.2: 232480312-232480312 on Build GRCh38
rs917483470 Chr.2: 232480273-232480273 on Build GRCh38
rs770543976 Chr.2: 232480414-232480414 on Build GRCh38
rs765282538 Chr.2: 232480286-232480286 on Build GRCh38
rs778036027 Chr.2: 232480400-232480400 on Build GRCh38
rs1048151553 Chr.2: 232480339-232480339 on Build GRCh38
rs954234186 Chr.2: 232480211-232480211 on Build GRCh38
rs751349206 Chr.2: 232480354-232480354 on Build GRCh38
rs192434358 Chr.2: 232480283-232480283 on Build GRCh38
rs587777129 Chr.2: 232480203-232480203 on Build GRCh38
rs531228026 Chr.2: 232480387-232480387 on Build GRCh38
rs200607176 Chr.2: 232480217-232480217 on Build GRCh38
rs780048518 Chr.2: 232480250-232480250 on Build GRCh38
rs372304234 Chr.2: 232480382-232480382 on Build GRCh38
rs779333975 Chr.2: 232480269-232480269 on Build GRCh38
rs776343913 Chr.2: 232480284-232480284 on Build GRCh38
rs1027424558 Chr.2: 232480239-232480239 on Build GRCh38
rs563275145 Chr.2: 232480401-232480401 on Build GRCh38
rs545152411 Chr.2: 232480305-232480305 on Build GRCh38
rs371645572 Chr.2: 232480234-232480234 on Build GRCh38
rs745379092 Chr.2: 232480420-232480420 on Build GRCh38
rs750573745 Chr.2: 232480237-232480237 on Build GRCh38
rs376978175 Chr.2: 232480422-232480422 on Build GRCh38
rs775505701 Chr.2: 232480289-232480289 on Build GRCh38
rs772938429 Chr.2: 232480280-232480280 on Build GRCh38
rs370047492 Chr.2: 232480385-232480385 on Build GRCh38
rs372181905 Chr.2: 232480276-232480276 on Build GRCh38
rs369256390 Chr.2: 232480350-232480350 on Build GRCh38
rs748515903 Chr.2: 232480275-232480275 on Build GRCh38
rs2741279 Chr.2: 232480281-232480281 on Build GRCh38
rs978851564 Chr.2: 232480214-232480214 on Build GRCh38
rs765265903 Chr.2: 232480205-232480205 on Build GRCh38
rs756331377 Chr.2: 232480372-232480372 on Build GRCh38
rs954066706 Chr.2: 232480344-232480344 on Build GRCh38

View Full Product Details