Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 617710-617965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR5
Entrez Gene ID
53841
Gene Name
cadherin related family member 5
Gene Aliases
MLPCDH, MU-PCDH, MUCDHL, MUPCDH
UniGene
Hs.165619
Gene Chromosome Location
Chr.11: 616565-625067 on Build GRCh38
Transcripts
NM_001171968.1 NM_021924.4 NM_031264.3 XM_006718253.3 XM_011520188.2 XM_011520189.2 XM_011520190.2 XM_011520191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671057 c.2150C>T p.A717V Chr.11: 617739-617739 on Build GRCh38
6431560 c.2148G>T p.Q716H Chr.11: 617741-617741 on Build GRCh38
1298288 c.2118G>A p.P706P Chr.11: 617954-617954 on Build GRCh38
5492722 c.2140G>A p.D714N Chr.11: 617749-617749 on Build GRCh38
6374904 c.2173A>G p.N725D Chr.11: 617716-617716 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs529527562 Chr.11: 617924-617924 on Build GRCh38
rs371175831 Chr.11: 617929-617929 on Build GRCh38
rs905116432 Chr.11: 617845-617845 on Build GRCh38
rs775863307 Chr.11: 617913-617913 on Build GRCh38
rs769876358 Chr.11: 617908-617908 on Build GRCh38
rs781589290 Chr.11: 617946-617946 on Build GRCh38
rs186765155 Chr.11: 617796-617796 on Build GRCh38
rs779552926 Chr.11: 617925-617925 on Build GRCh38
rs909932733 Chr.11: 617802-617802 on Build GRCh38
rs552855718 Chr.11: 617855-617855 on Build GRCh38
rs756124038 Chr.11: 617771-617771 on Build GRCh38
rs1055838699 Chr.11: 617810-617810 on Build GRCh38
rs111246420 Chr.11: 617854-617854 on Build GRCh38
rs544753589 Chr.11: 617883-617883 on Build GRCh38
rs372636570 Chr.11: 617791-617791 on Build GRCh38
rs957715577 Chr.11: 617770-617770 on Build GRCh38
rs369792192 Chr.11: 617886-617886 on Build GRCh38
rs12385783 Chr.11: 617780-617780 on Build GRCh38
rs893518822 Chr.11: 617866-617866 on Build GRCh38
rs1046282436 Chr.11: 617853-617853 on Build GRCh38
rs543765073 Chr.11: 617917-617917 on Build GRCh38
rs573809635 Chr.11: 617844-617844 on Build GRCh38
rs776893242 Chr.11: 617904-617904 on Build GRCh38
rs982877206 Chr.11: 617797-617797 on Build GRCh38
rs750040345 Chr.11: 617923-617923 on Build GRCh38
rs1015376034 Chr.11: 617887-617887 on Build GRCh38
rs182907729 Chr.11: 617874-617874 on Build GRCh38
rs534793497 Chr.11: 617741-617741 on Build GRCh38
rs201360349 Chr.11: 617738-617738 on Build GRCh38
rs369213863 Chr.11: 617785-617785 on Build GRCh38
rs571665990 Chr.11: 617784-617784 on Build GRCh38
rs1011072481 Chr.11: 617872-617872 on Build GRCh38
rs754933229 Chr.11: 617930-617930 on Build GRCh38
rs922787873 Chr.11: 617790-617790 on Build GRCh38
rs951532187 Chr.11: 617898-617898 on Build GRCh38
rs746033707 Chr.11: 617947-617947 on Build GRCh38
rs908269069 Chr.11: 617727-617727 on Build GRCh38
rs763459526 Chr.11: 617914-617914 on Build GRCh38
rs751913571 Chr.11: 617918-617918 on Build GRCh38
rs962400345 Chr.11: 617897-617897 on Build GRCh38
rs779874989 Chr.11: 617778-617778 on Build GRCh38
rs768342805 Chr.11: 617779-617779 on Build GRCh38
rs920905321 Chr.11: 617902-617902 on Build GRCh38
rs111307862 Chr.11: 617833-617833 on Build GRCh38
rs199885960 Chr.11: 617740-617740 on Build GRCh38
rs899903005 Chr.11: 617801-617801 on Build GRCh38
rs776262999 Chr.11: 617882-617883 on Build GRCh38
rs915939097 Chr.11: 617931-617931 on Build GRCh38
rs1030685973 Chr.11: 617761-617761 on Build GRCh38
rs941360953 Chr.11: 617808-617808 on Build GRCh38
rs574882718 Chr.11: 617901-617901 on Build GRCh38
rs1052145423 Chr.11: 617859-617859 on Build GRCh38
rs759514494 Chr.11: 617905-617905 on Build GRCh38
rs747921583 Chr.11: 617939-617939 on Build GRCh38
rs1050571021 Chr.11: 617786-617786 on Build GRCh38
rs759105356 Chr.11: 617903-617904 on Build GRCh38
rs560553793 Chr.11: 617919-617919 on Build GRCh38
rs756889133 Chr.11: 617739-617739 on Build GRCh38
rs964028768 Chr.11: 617875-617875 on Build GRCh38
rs771274750 Chr.11: 617818-617818 on Build GRCh38
rs771330067 Chr.11: 617941-617941 on Build GRCh38

View Full Product Details