Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73232968-73233195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149B1
Entrez Gene ID
317662
Gene Name
family with sequence similarity 149 member B1
Gene Aliases
KIAA0974
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73168090-73242181 on Build GRCh38
Transcripts
NM_173348.1 XM_005269744.2 XM_005269745.2 XM_005269747.3 XM_011539737.2 XM_017016164.1 XM_017016165.1 XM_017016166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551046 c.1183C>T p.R395* Chr.10: 73232994-73232994 on Build GRCh38
2161820 c.1167T>C p.S389S Chr.10: 73232978-73232978 on Build GRCh38
5639291 c.1159G>A p.G387R Chr.10: 73232970-73232970 on Build GRCh38
4856702 c.1340G>A p.R447K Chr.10: 73233151-73233151 on Build GRCh38
7348546 c.1271G>A p.R424Q Chr.10: 73233082-73233082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144804269 Chr.10: 73232994-73232994 on Build GRCh38
rs771923238 Chr.10: 73233157-73233157 on Build GRCh38
rs531266047 Chr.10: 73233082-73233082 on Build GRCh38
rs775465672 Chr.10: 73233048-73233048 on Build GRCh38
rs926129957 Chr.10: 73233143-73233143 on Build GRCh38
rs776595563 Chr.10: 73233093-73233093 on Build GRCh38
rs369028725 Chr.10: 73233018-73233018 on Build GRCh38
rs371485920 Chr.10: 73233107-73233107 on Build GRCh38
rs968366251 Chr.10: 73233067-73233067 on Build GRCh38
rs983908349 Chr.10: 73233023-73233023 on Build GRCh38
rs200426792 Chr.10: 73233105-73233105 on Build GRCh38
rs993432121 Chr.10: 73233092-73233092 on Build GRCh38
rs767987832 Chr.10: 73233101-73233101 on Build GRCh38
rs542802146 Chr.10: 73233010-73233010 on Build GRCh38
rs1039766170 Chr.10: 73233164-73233164 on Build GRCh38
rs772011416 Chr.10: 73233039-73233039 on Build GRCh38
rs1009856196 Chr.10: 73233063-73233063 on Build GRCh38
rs567292300 Chr.10: 73233094-73233094 on Build GRCh38
rs202136724 Chr.10: 73233000-73233000 on Build GRCh38
rs770568936 Chr.10: 73233004-73233004 on Build GRCh38
rs376018524 Chr.10: 73233081-73233081 on Build GRCh38
rs753294444 Chr.10: 73233102-73233102 on Build GRCh38
rs147920257 Chr.10: 73233156-73233156 on Build GRCh38
rs951645895 Chr.10: 73233095-73233095 on Build GRCh38
rs760240891 Chr.10: 73233007-73233007 on Build GRCh38
rs756665429 Chr.10: 73233103-73233103 on Build GRCh38
rs1053170465 Chr.10: 73233159-73233159 on Build GRCh38
rs759437374 Chr.10: 73233113-73233113 on Build GRCh38
rs764971459 Chr.10: 73233115-73233115 on Build GRCh38
rs767913197 Chr.10: 73233109-73233109 on Build GRCh38
rs370597793 Chr.10: 73233085-73233085 on Build GRCh38
rs372801139 Chr.10: 73233066-73233066 on Build GRCh38
rs373653360 Chr.10: 73233086-73233086 on Build GRCh38
rs748791406 Chr.10: 73233104-73233104 on Build GRCh38
rs563849642 Chr.10: 73232995-73232995 on Build GRCh38

View Full Product Details