Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1839266-1839493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
UniGene
Hs.729604
Gene Chromosome Location
Chr.16: 1833983-1872178 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7376914 c.321T>G p.I107M Chr.16: 1839420-1839420 on Build GRCh38
7315904 c.466G>A p.E156K Chr.16: 1839275-1839275 on Build GRCh38
7376913 c.1053T>G p.I351M Chr.16: 1839420-1839420 on Build GRCh38
4058818 c.1107T>G p.F369L Chr.16: 1839366-1839366 on Build GRCh38
1478581 c.312T>C p.C104C Chr.16: 1839429-1839429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747236111 Chr.16: 1839336-1839336 on Build GRCh38
rs779513355 Chr.16: 1839400-1839401 on Build GRCh38
rs909331976 Chr.16: 1839363-1839363 on Build GRCh38
rs758723085 Chr.16: 1839466-1839466 on Build GRCh38
rs755596200 Chr.16: 1839311-1839311 on Build GRCh38
rs748509110 Chr.16: 1839316-1839316 on Build GRCh38
rs749802810 Chr.16: 1839310-1839310 on Build GRCh38
rs751791205 Chr.16: 1839351-1839354 on Build GRCh38
rs145222212 Chr.16: 1839366-1839366 on Build GRCh38
rs751186846 Chr.16: 1839414-1839414 on Build GRCh38
rs766633975 Chr.16: 1839330-1839333 on Build GRCh38
rs143531383 Chr.16: 1839420-1839420 on Build GRCh38
rs770995208 Chr.16: 1839351-1839351 on Build GRCh38
rs985183192 Chr.16: 1839362-1839362 on Build GRCh38
rs377458120 Chr.16: 1839318-1839318 on Build GRCh38
rs751190600 Chr.16: 1839293-1839293 on Build GRCh38
rs370800787 Chr.16: 1839330-1839330 on Build GRCh38
rs781620229 Chr.16: 1839449-1839454 on Build GRCh38
rs777438650 Chr.16: 1839287-1839287 on Build GRCh38
rs933928784 Chr.16: 1839449-1839449 on Build GRCh38
rs779428308 Chr.16: 1839458-1839458 on Build GRCh38
rs779119124 Chr.16: 1839313-1839313 on Build GRCh38
rs972548876 Chr.16: 1839439-1839439 on Build GRCh38
rs755719433 Chr.16: 1839445-1839445 on Build GRCh38
rs201609657 Chr.16: 1839309-1839309 on Build GRCh38
rs763807926 Chr.16: 1839401-1839401 on Build GRCh38
rs776748763 Chr.16: 1839360-1839360 on Build GRCh38
rs376572512 Chr.16: 1839361-1839361 on Build GRCh38
rs766989673 Chr.16: 1839447-1839447 on Build GRCh38
rs567019697 Chr.16: 1839424-1839424 on Build GRCh38
rs149166943 Chr.16: 1839391-1839391 on Build GRCh38
rs748618534 Chr.16: 1839460-1839460 on Build GRCh38
rs775319534 Chr.16: 1839385-1839385 on Build GRCh38
rs200721888 Chr.16: 1839395-1839395 on Build GRCh38
rs202223532 Chr.16: 1839467-1839467 on Build GRCh38
rs971645189 Chr.16: 1839294-1839294 on Build GRCh38

View Full Product Details