Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9344441-9344672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
UniGene
Hs.555902
Gene Chromosome Location
Chr.2: 9206765-9405683 on Build GRCh38
Transcripts
NM_001135191.1 NM_003887.2 XM_006711898.1 XM_006711901.3 XM_006711902.1 XM_011510403.2 XM_011510404.2 XM_011510405.2 XM_011510406.2 XM_011510407.2 XM_011510408.2 XM_011510409.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761582 c.901A>G p.N301D Chr.2: 9344583-9344583 on Build GRCh38
1023643 c.862C>T p.R288C Chr.2: 9344544-9344544 on Build GRCh38
7078435 c.901A>T p.N301Y Chr.2: 9344583-9344583 on Build GRCh38
5518274 c.951C>T p.D317D Chr.2: 9344633-9344633 on Build GRCh38
180342 c.922C>T p.R308W Chr.2: 9344604-9344604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751479586 Chr.2: 9344515-9344515 on Build GRCh38
rs1056380934 Chr.2: 9344597-9344597 on Build GRCh38
rs759370018 Chr.2: 9344516-9344516 on Build GRCh38
rs769838169 Chr.2: 9344620-9344620 on Build GRCh38
rs760405024 Chr.2: 9344644-9344644 on Build GRCh38
rs763602369 Chr.2: 9344645-9344645 on Build GRCh38
rs753245751 Chr.2: 9344545-9344545 on Build GRCh38
rs776513072 Chr.2: 9344505-9344505 on Build GRCh38
rs761935035 Chr.2: 9344610-9344610 on Build GRCh38
rs780267210 Chr.2: 9344600-9344600 on Build GRCh38
rs753508196 Chr.2: 9344589-9344589 on Build GRCh38
rs757858736 Chr.2: 9344559-9344559 on Build GRCh38
rs905087721 Chr.2: 9344561-9344561 on Build GRCh38
rs753506040 Chr.2: 9344647-9344647 on Build GRCh38
rs747266776 Chr.2: 9344604-9344604 on Build GRCh38
rs767296380 Chr.2: 9344523-9344523 on Build GRCh38
rs779611839 Chr.2: 9344569-9344569 on Build GRCh38
rs777343307 Chr.2: 9344541-9344541 on Build GRCh38
rs576089706 Chr.2: 9344633-9344633 on Build GRCh38
rs755665966 Chr.2: 9344540-9344540 on Build GRCh38
rs762982469 Chr.2: 9344507-9344507 on Build GRCh38
rs905113318 Chr.2: 9344513-9344513 on Build GRCh38
rs1020353303 Chr.2: 9344535-9344535 on Build GRCh38
rs746342296 Chr.2: 9344574-9344574 on Build GRCh38
rs1032601093 Chr.2: 9344520-9344520 on Build GRCh38
rs768834729 Chr.2: 9344500-9344500 on Build GRCh38
rs536801863 Chr.2: 9344495-9344495 on Build GRCh38
rs374692643 Chr.2: 9344498-9344498 on Build GRCh38
rs559656538 Chr.2: 9344487-9344487 on Build GRCh38
rs201120621 Chr.2: 9344514-9344514 on Build GRCh38
rs1045006619 Chr.2: 9344555-9344555 on Build GRCh38
rs774339512 Chr.2: 9344621-9344621 on Build GRCh38
rs768678377 Chr.2: 9344534-9344534 on Build GRCh38
rs774456784 Chr.2: 9344640-9344640 on Build GRCh38
rs776797223 Chr.2: 9344609-9344609 on Build GRCh38
rs772540793 Chr.2: 9344494-9344494 on Build GRCh38
rs772628333 Chr.2: 9344593-9344593 on Build GRCh38
rs768723108 Chr.2: 9344605-9344605 on Build GRCh38

View Full Product Details