Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14505564-14505817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
UniGene
Hs.517819
Gene Chromosome Location
Chr.3: 14489111-14602996 on Build GRCh38
Transcripts
NM_001080423.3 XM_011534141.2 XM_011534142.2 XM_011534143.2 XM_011534145.2 XM_011534146.2

SNP Detail

SNP ID Chromosome Location
rs755177908 Chr.3: 14505793-14505793 on Build GRCh38
rs776686930 Chr.3: 14505761-14505761 on Build GRCh38
rs760138571 Chr.3: 14505700-14505700 on Build GRCh38
rs760584263 Chr.3: 14505649-14505649 on Build GRCh38
rs780256510 Chr.3: 14505676-14505676 on Build GRCh38
rs183031950 Chr.3: 14505743-14505743 on Build GRCh38
rs375348449 Chr.3: 14505697-14505697 on Build GRCh38
rs769314002 Chr.3: 14505681-14505681 on Build GRCh38
rs377452768 Chr.3: 14505623-14505623 on Build GRCh38
rs781747884 Chr.3: 14505684-14505684 on Build GRCh38
rs371768125 Chr.3: 14505755-14505755 on Build GRCh38
rs1014889487 Chr.3: 14505758-14505758 on Build GRCh38
rs747556022 Chr.3: 14505680-14505680 on Build GRCh38
rs764032337 Chr.3: 14505702-14505702 on Build GRCh38
rs905642878 Chr.3: 14505715-14505715 on Build GRCh38
rs372323676 Chr.3: 14505696-14505696 on Build GRCh38
rs547662300 Chr.3: 14505704-14505704 on Build GRCh38
rs766665902 Chr.3: 14505722-14505722 on Build GRCh38
rs753650139 Chr.3: 14505703-14505703 on Build GRCh38
rs534745511 Chr.3: 14505624-14505624 on Build GRCh38
rs745574318 Chr.3: 14505753-14505753 on Build GRCh38
rs752690741 Chr.3: 14505638-14505638 on Build GRCh38
rs973262839 Chr.3: 14505778-14505778 on Build GRCh38
rs772908070 Chr.3: 14505777-14505777 on Build GRCh38
rs765287102 Chr.3: 14505635-14505636 on Build GRCh38
rs773819845 Chr.3: 14505634-14505634 on Build GRCh38
rs770642884 Chr.3: 14505632-14505632 on Build GRCh38
rs769189397 Chr.3: 14505619-14505619 on Build GRCh38
rs867921988 Chr.3: 14505678-14505678 on Build GRCh38
rs866579868 Chr.3: 14505750-14505750 on Build GRCh38
rs750704880 Chr.3: 14505705-14505705 on Build GRCh38
rs536443338 Chr.3: 14505781-14505781 on Build GRCh38
rs371589692 Chr.3: 14505665-14505665 on Build GRCh38
rs767400239 Chr.3: 14505594-14505604 on Build GRCh38
rs914797283 Chr.3: 14505780-14505780 on Build GRCh38
rs377608784 Chr.3: 14505597-14505597 on Build GRCh38
rs751769776 Chr.3: 14505786-14505786 on Build GRCh38
rs375975446 Chr.3: 14505610-14505610 on Build GRCh38
rs757372923 Chr.3: 14505636-14505638 on Build GRCh38
rs778953618 Chr.3: 14505596-14505596 on Build GRCh38
rs753655527 Chr.3: 14505654-14505654 on Build GRCh38
rs757470678 Chr.3: 14505594-14505594 on Build GRCh38
rs200381587 Chr.3: 14505664-14505664 on Build GRCh38
rs758659394 Chr.3: 14505711-14505711 on Build GRCh38
rs371332318 Chr.3: 14505598-14505598 on Build GRCh38
rs759099315 Chr.3: 14505691-14505691 on Build GRCh38
rs768148947 Chr.3: 14505756-14505756 on Build GRCh38
rs748562478 Chr.3: 14505686-14505686 on Build GRCh38
rs755055241 Chr.3: 14505742-14505742 on Build GRCh38
rs748600380 Chr.3: 14505748-14505748 on Build GRCh38
rs374795024 Chr.3: 14505762-14505762 on Build GRCh38
rs750604260 Chr.3: 14505673-14505673 on Build GRCh38
rs370222047 Chr.3: 14505635-14505635 on Build GRCh38
rs921994252 Chr.3: 14505798-14505798 on Build GRCh38
rs766725365 Chr.3: 14505784-14505784 on Build GRCh38
rs765118255 Chr.3: 14505763-14505763 on Build GRCh38
rs61731938 Chr.3: 14505674-14505674 on Build GRCh38
rs374133419 Chr.3: 14505637-14505637 on Build GRCh38
rs369067748 Chr.3: 14505752-14505752 on Build GRCh38
rs532562302 Chr.3: 14505689-14505689 on Build GRCh38
rs751708388 Chr.3: 14505741-14505741 on Build GRCh38
rs61731939 Chr.3: 14505690-14505690 on Build GRCh38
rs747503091 Chr.3: 14505618-14505618 on Build GRCh38
rs752596660 Chr.3: 14505608-14505609 on Build GRCh38
rs1001584119 Chr.3: 14505720-14505720 on Build GRCh38

View Full Product Details