Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44080274-44080527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC5
Entrez Gene ID
10014
Gene Name
histone deacetylase 5
Gene Aliases
HD5, NY-CO-9
UniGene
Hs.438782
Gene Chromosome Location
Chr.17: 44076753-44123646 on Build GRCh38
Transcripts
NM_001015053.1 NM_005474.4 XM_005256905.1 XM_005256906.4 XM_011524149.1 XM_011524150.1 XM_017023988.1 XM_017023989.1 XM_017023990.1 XM_017023991.1 XM_017023992.1 XM_017023993.1 XM_017023994.1 XM_017023995.1 XM_017023996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6565630 c.2825+2T>G p.? Chr.17: 44080399-44080399 on Build GRCh38
4753673 c.2746G>A p.V916M Chr.17: 44080480-44080480 on Build GRCh38
3730023 c.2793_2794insC p.I932fs*24 Chr.17: 44080432-44080433 on Build GRCh38
1383594 c.2793delC p.I932fs*14 Chr.17: 44080433-44080433 on Build GRCh38
4539905 c.2750G>A p.G917E Chr.17: 44080476-44080476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749856924 Chr.17: 44080365-44080365 on Build GRCh38
rs550032880 Chr.17: 44080328-44080328 on Build GRCh38
rs780519704 Chr.17: 44080353-44080353 on Build GRCh38
rs139019058 Chr.17: 44080463-44080463 on Build GRCh38
rs747263506 Chr.17: 44080490-44080490 on Build GRCh38
rs192218780 Chr.17: 44080423-44080423 on Build GRCh38
rs765280219 Chr.17: 44080419-44080419 on Build GRCh38
rs755670175 Chr.17: 44080312-44080312 on Build GRCh38
rs759409556 Chr.17: 44080505-44080505 on Build GRCh38
rs897736763 Chr.17: 44080412-44080412 on Build GRCh38
rs756471886 Chr.17: 44080439-44080439 on Build GRCh38
rs766315501 Chr.17: 44080406-44080406 on Build GRCh38
rs773450329 Chr.17: 44080485-44080485 on Build GRCh38
rs535899624 Chr.17: 44080391-44080391 on Build GRCh38
rs753824674 Chr.17: 44080410-44080410 on Build GRCh38
rs751222972 Chr.17: 44080432-44080432 on Build GRCh38
rs369361035 Chr.17: 44080480-44080480 on Build GRCh38
rs901599101 Chr.17: 44080308-44080308 on Build GRCh38
rs770174487 Chr.17: 44080432-44080433 on Build GRCh38
rs1038175589 Chr.17: 44080362-44080362 on Build GRCh38
rs774353556 Chr.17: 44080372-44080372 on Build GRCh38
rs990946311 Chr.17: 44080497-44080497 on Build GRCh38
rs760494828 Chr.17: 44080398-44080398 on Build GRCh38
rs749663119 Chr.17: 44080452-44080452 on Build GRCh38
rs139098243 Chr.17: 44080424-44080424 on Build GRCh38
rs780452318 Chr.17: 44080441-44080441 on Build GRCh38
rs752185350 Chr.17: 44080421-44080421 on Build GRCh38
rs35610404 Chr.17: 44080336-44080336 on Build GRCh38
rs748208936 Chr.17: 44080481-44080481 on Build GRCh38
rs772296546 Chr.17: 44080387-44080387 on Build GRCh38
rs754901311 Chr.17: 44080450-44080450 on Build GRCh38
rs537284972 Chr.17: 44080340-44080340 on Build GRCh38
rs776511917 Chr.17: 44080504-44080504 on Build GRCh38
rs770837519 Chr.17: 44080501-44080501 on Build GRCh38
rs993908235 Chr.17: 44080390-44080390 on Build GRCh38
rs850856 Chr.17: 44080371-44080371 on Build GRCh38
rs367647540 Chr.17: 44080341-44080341 on Build GRCh38
rs759605807 Chr.17: 44080415-44080415 on Build GRCh38

View Full Product Details