Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41237829-41238088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001278599.1 NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855871 c.820G>A p.G274R Chr.19: 41237980-41237980 on Build GRCh38
5798833 c.901C>T p.P301S Chr.19: 41238061-41238061 on Build GRCh38
1481109 c.867_868insC p.H292fs*47 Chr.19: 41238027-41238028 on Build GRCh38
6947522 c.889G>A p.G297S Chr.19: 41238049-41238049 on Build GRCh38
4560394 c.841G>A p.E281K Chr.19: 41238001-41238001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893777534 Chr.19: 41237907-41237907 on Build GRCh38
rs757185500 Chr.19: 41237964-41237964 on Build GRCh38
rs7249222 Chr.19: 41237956-41237956 on Build GRCh38
rs1032113003 Chr.19: 41237997-41237997 on Build GRCh38
rs141302305 Chr.19: 41238025-41238025 on Build GRCh38
rs568452639 Chr.19: 41238001-41238001 on Build GRCh38
rs929326745 Chr.19: 41237941-41237941 on Build GRCh38
rs763605285 Chr.19: 41237958-41237958 on Build GRCh38
rs755029075 Chr.19: 41237991-41237991 on Build GRCh38
rs113249799 Chr.19: 41238024-41238024 on Build GRCh38
rs773696483 Chr.19: 41237939-41237939 on Build GRCh38
rs768649416 Chr.19: 41237915-41237915 on Build GRCh38
rs748294158 Chr.19: 41237992-41237992 on Build GRCh38
rs770488015 Chr.19: 41237918-41237918 on Build GRCh38
rs61737383 Chr.19: 41237979-41237979 on Build GRCh38
rs749768475 Chr.19: 41238066-41238066 on Build GRCh38
rs200703161 Chr.19: 41237936-41237936 on Build GRCh38
rs777230616 Chr.19: 41237917-41237917 on Build GRCh38
rs533084444 Chr.19: 41237906-41237906 on Build GRCh38
rs1061079 Chr.19: 41238067-41238067 on Build GRCh38
rs751738506 Chr.19: 41238043-41238043 on Build GRCh38
rs199988157 Chr.19: 41238044-41238044 on Build GRCh38
rs760246352 Chr.19: 41238015-41238015 on Build GRCh38
rs758581229 Chr.19: 41237990-41237991 on Build GRCh38
rs771258043 Chr.19: 41238068-41238068 on Build GRCh38
rs748208134 Chr.19: 41238054-41238054 on Build GRCh38
rs191926606 Chr.19: 41238005-41238005 on Build GRCh38
rs201003955 Chr.19: 41238014-41238014 on Build GRCh38
rs35465736 Chr.19: 41237943-41237943 on Build GRCh38
rs778687509 Chr.19: 41237904-41237904 on Build GRCh38
rs551157335 Chr.19: 41237913-41237913 on Build GRCh38
rs780193824 Chr.19: 41237911-41237911 on Build GRCh38
rs778012871 Chr.19: 41238027-41238028 on Build GRCh38
rs374699228 Chr.19: 41237978-41237978 on Build GRCh38
rs767166443 Chr.19: 41237945-41237945 on Build GRCh38
rs745732524 Chr.19: 41237905-41237905 on Build GRCh38
rs753706427 Chr.19: 41237959-41237959 on Build GRCh38
rs910412418 Chr.19: 41238031-41238031 on Build GRCh38
rs747220380 Chr.19: 41237981-41237981 on Build GRCh38
rs780282463 Chr.19: 41237980-41237980 on Build GRCh38
rs765219468 Chr.19: 41237973-41237973 on Build GRCh38
rs370447193 Chr.19: 41238020-41238020 on Build GRCh38
rs201596308 Chr.19: 41238061-41238061 on Build GRCh38
rs973173615 Chr.19: 41238045-41238045 on Build GRCh38
rs778289997 Chr.19: 41238065-41238065 on Build GRCh38
rs773894800 Chr.19: 41237998-41237998 on Build GRCh38
rs758164020 Chr.19: 41238032-41238032 on Build GRCh38
rs777643850 Chr.19: 41237896-41237896 on Build GRCh38
rs774572664 Chr.19: 41238069-41238069 on Build GRCh38
rs756985286 Chr.19: 41237899-41237899 on Build GRCh38
rs112094962 Chr.19: 41238047-41238047 on Build GRCh38
rs1027287501 Chr.19: 41237994-41237994 on Build GRCh38
rs796712105 Chr.19: 41237967-41237967 on Build GRCh38
rs976334285 Chr.19: 41238050-41238050 on Build GRCh38
rs770402054 Chr.19: 41237996-41237996 on Build GRCh38
rs775032851 Chr.19: 41237952-41237952 on Build GRCh38
rs750359676 Chr.19: 41238029-41238029 on Build GRCh38
rs1037765875 Chr.19: 41237857-41237857 on Build GRCh38
rs753792668 Chr.19: 41237897-41237897 on Build GRCh38
rs781611976 Chr.19: 41238053-41238053 on Build GRCh38
rs139491068 Chr.19: 41237944-41237944 on Build GRCh38

View Full Product Details