Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103520303-103520538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKB
Entrez Gene ID
1152
Gene Name
creatine kinase B
Gene Aliases
B-CK, BCK, CKBB, HEL-211, HEL-S-29
UniGene
Hs.173724
Gene Chromosome Location
Chr.14: 103519658-103522859 on Build GRCh38
Transcripts
NM_001823.4 XM_017020951.1 XM_017020952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2025552 c.731delG p.G244fs*3 Chr.14: 103520515-103520515 on Build GRCh38
7284086 c.778-8G>A p.? Chr.14: 103520319-103520319 on Build GRCh38
5965680 c.723G>A p.Q241Q Chr.14: 103520523-103520523 on Build GRCh38
3936424 c.778-2A>T p.? Chr.14: 103520313-103520313 on Build GRCh38
4167720 c.755G>T p.R252L Chr.14: 103520491-103520491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761370221 Chr.14: 103520500-103520500 on Build GRCh38
rs971346285 Chr.14: 103520432-103520432 on Build GRCh38
rs780082737 Chr.14: 103520441-103520441 on Build GRCh38
rs768949165 Chr.14: 103520498-103520499 on Build GRCh38
rs752830110 Chr.14: 103520442-103520442 on Build GRCh38
rs369244250 Chr.14: 103520436-103520436 on Build GRCh38
rs781402849 Chr.14: 103520472-103520472 on Build GRCh38
rs767405482 Chr.14: 103520517-103520517 on Build GRCh38
rs998965472 Chr.14: 103520375-103520375 on Build GRCh38
rs373166125 Chr.14: 103520439-103520439 on Build GRCh38
rs776935018 Chr.14: 103520401-103520401 on Build GRCh38
rs768452864 Chr.14: 103520360-103520360 on Build GRCh38
rs775905075 Chr.14: 103520481-103520481 on Build GRCh38
rs369370975 Chr.14: 103520502-103520502 on Build GRCh38
rs377598922 Chr.14: 103520492-103520492 on Build GRCh38
rs542400698 Chr.14: 103520460-103520460 on Build GRCh38
rs774966043 Chr.14: 103520328-103520328 on Build GRCh38
rs746442711 Chr.14: 103520475-103520475 on Build GRCh38
rs573690664 Chr.14: 103520453-103520453 on Build GRCh38
rs113915995 Chr.14: 103520468-103520468 on Build GRCh38
rs758511225 Chr.14: 103520443-103520443 on Build GRCh38
rs944129155 Chr.14: 103520484-103520484 on Build GRCh38
rs762387560 Chr.14: 103520329-103520329 on Build GRCh38
rs762342435 Chr.14: 103520427-103520427 on Build GRCh38
rs763737882 Chr.14: 103520330-103520330 on Build GRCh38
rs955669368 Chr.14: 103520364-103520364 on Build GRCh38
rs767189152 Chr.14: 103520428-103520428 on Build GRCh38
rs774095172 Chr.14: 103520493-103520493 on Build GRCh38
rs368946669 Chr.14: 103520368-103520369 on Build GRCh38
rs984540171 Chr.14: 103520464-103520464 on Build GRCh38
rs914799971 Chr.14: 103520409-103520409 on Build GRCh38
rs148243589 Chr.14: 103520505-103520505 on Build GRCh38
rs957222903 Chr.14: 103520402-103520402 on Build GRCh38
rs762475613 Chr.14: 103520491-103520491 on Build GRCh38
rs375018448 Chr.14: 103520418-103520418 on Build GRCh38
rs1033078926 Chr.14: 103520393-103520393 on Build GRCh38
rs751127227 Chr.14: 103520337-103520337 on Build GRCh38
rs1023179903 Chr.14: 103520424-103520424 on Build GRCh38
rs762330719 Chr.14: 103520344-103520344 on Build GRCh38
rs990383162 Chr.14: 103520392-103520392 on Build GRCh38
rs771366122 Chr.14: 103520461-103520461 on Build GRCh38
rs770294318 Chr.14: 103520480-103520480 on Build GRCh38
rs113078547 Chr.14: 103520429-103520429 on Build GRCh38

View Full Product Details