Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32166337-32166588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL8
Entrez Gene ID
80864
Gene Name
EGF like domain multiple 8
Gene Aliases
C6orf8, NG3
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32164605-32168285 on Build GRCh38
Transcripts
NM_030652.3 NR_037860.1 NR_037861.1

Gene Symbol
PPT2-EGFL8
Entrez Gene ID
100532746
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32153999-32168285 on Build GRCh38
Transcripts
NM_030652.3 NR_037860.1 NR_037861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652185 c.170T>C p.V57A Chr.6: 32166566-32166566 on Build GRCh38
5870292 c.102-1G>A p.? Chr.6: 32166497-32166497 on Build GRCh38
6193247 c.117C>T p.S39S Chr.6: 32166513-32166513 on Build GRCh38
6754695 c.102-4G>A p.? Chr.6: 32166494-32166494 on Build GRCh38
6652181 c.151G>A p.E51K Chr.6: 32166547-32166547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768104469 Chr.6: 32166517-32166517 on Build GRCh38
rs771976535 Chr.6: 32166462-32166462 on Build GRCh38
rs774671618 Chr.6: 32166542-32166544 on Build GRCh38
rs72289190 Chr.6: 32166367-32166373 on Build GRCh38
rs575900965 Chr.6: 32166534-32166534 on Build GRCh38
rs35319377 Chr.6: 32166500-32166501 on Build GRCh38
rs972473394 Chr.6: 32166480-32166480 on Build GRCh38
rs1031263305 Chr.6: 32166427-32166427 on Build GRCh38
rs567711648 Chr.6: 32166494-32166494 on Build GRCh38
rs1012906843 Chr.6: 32166397-32166397 on Build GRCh38
rs953689697 Chr.6: 32166424-32166424 on Build GRCh38
rs1045677126 Chr.6: 32166399-32166399 on Build GRCh38
rs769920311 Chr.6: 32166493-32166493 on Build GRCh38
rs377454015 Chr.6: 32166547-32166547 on Build GRCh38
rs762314805 Chr.6: 32166504-32166504 on Build GRCh38
rs148110134 Chr.6: 32166518-32166518 on Build GRCh38
rs753463061 Chr.6: 32166525-32166525 on Build GRCh38
rs752459687 Chr.6: 32166546-32166546 on Build GRCh38
rs748037459 Chr.6: 32166460-32166460 on Build GRCh38
rs965091448 Chr.6: 32166370-32166370 on Build GRCh38
rs745434046 Chr.6: 32166503-32166503 on Build GRCh38
rs919294715 Chr.6: 32166510-32166510 on Build GRCh38
rs778719150 Chr.6: 32166543-32166543 on Build GRCh38
rs555877656 Chr.6: 32166402-32166402 on Build GRCh38
rs375116850 Chr.6: 32166523-32166523 on Build GRCh38
rs769849513 Chr.6: 32166566-32166566 on Build GRCh38
rs143726348 Chr.6: 32166497-32166497 on Build GRCh38
rs778539680 Chr.6: 32166436-32166436 on Build GRCh38
rs746963804 Chr.6: 32166564-32166564 on Build GRCh38
rs998924125 Chr.6: 32166405-32166405 on Build GRCh38
rs545453429 Chr.6: 32166542-32166542 on Build GRCh38
rs1011136450 Chr.6: 32166466-32166466 on Build GRCh38
rs909442410 Chr.6: 32166456-32166456 on Build GRCh38
rs764363570 Chr.6: 32166440-32166440 on Build GRCh38
rs773176213 Chr.6: 32166481-32166481 on Build GRCh38
rs997896942 Chr.6: 32166383-32166383 on Build GRCh38
rs765820216 Chr.6: 32166521-32166521 on Build GRCh38

View Full Product Details