Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55846438-55846697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES5A
Entrez Gene ID
221223
Gene Name
carboxylesterase 5A
Gene Aliases
CAUXIN, CES4C1, CES5, CES7, HEL126
UniGene
Hs.350800
Gene Chromosome Location
Chr.16: 55846154-55956031 on Build GRCh38
Transcripts
NM_001143685.1 NM_001190158.1 NM_145024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6851886 c.1535T>C p.F512S Chr.16: 55846494-55846494 on Build GRCh38
6144797 c.1785C>A p.L595L Chr.16: 55846481-55846481 on Build GRCh38
6851885 c.1772T>C p.F591S Chr.16: 55846494-55846494 on Build GRCh38
4061306 c.1451C>T p.P484L Chr.16: 55846578-55846578 on Build GRCh38
3754936 c.1644C>G p.L548L Chr.16: 55846622-55846622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760544900 Chr.16: 55846648-55846648 on Build GRCh38
rs755039115 Chr.16: 55846503-55846503 on Build GRCh38
rs556065376 Chr.16: 55846597-55846597 on Build GRCh38
rs575240574 Chr.16: 55846467-55846467 on Build GRCh38
rs1006004877 Chr.16: 55846518-55846518 on Build GRCh38
rs767548249 Chr.16: 55846551-55846551 on Build GRCh38
rs758064010 Chr.16: 55846545-55846545 on Build GRCh38
rs958211764 Chr.16: 55846649-55846649 on Build GRCh38
rs146255658 Chr.16: 55846547-55846547 on Build GRCh38
rs779176437 Chr.16: 55846587-55846587 on Build GRCh38
rs746504097 Chr.16: 55846634-55846636 on Build GRCh38
rs558363968 Chr.16: 55846506-55846506 on Build GRCh38
rs11860456 Chr.16: 55846568-55846568 on Build GRCh38
rs747381459 Chr.16: 55846528-55846528 on Build GRCh38
rs748591331 Chr.16: 55846515-55846515 on Build GRCh38
rs141258202 Chr.16: 55846499-55846499 on Build GRCh38
rs186119494 Chr.16: 55846667-55846667 on Build GRCh38
rs768498192 Chr.16: 55846640-55846641 on Build GRCh38
rs750375972 Chr.16: 55846562-55846562 on Build GRCh38
rs988245292 Chr.16: 55846654-55846654 on Build GRCh38
rs768221015 Chr.16: 55846535-55846535 on Build GRCh38
rs762533167 Chr.16: 55846533-55846533 on Build GRCh38
rs368968057 Chr.16: 55846647-55846647 on Build GRCh38
rs1032950826 Chr.16: 55846633-55846633 on Build GRCh38
rs374558958 Chr.16: 55846517-55846517 on Build GRCh38
rs760480313 Chr.16: 55846567-55846567 on Build GRCh38
rs771236888 Chr.16: 55846531-55846531 on Build GRCh38
rs539590082 Chr.16: 55846477-55846477 on Build GRCh38
rs775164933 Chr.16: 55846532-55846532 on Build GRCh38
rs781421042 Chr.16: 55846621-55846621 on Build GRCh38
rs779469867 Chr.16: 55846564-55846564 on Build GRCh38
rs774002308 Chr.16: 55846540-55846540 on Build GRCh38
rs373627529 Chr.16: 55846554-55846554 on Build GRCh38
rs761384775 Chr.16: 55846637-55846637 on Build GRCh38
rs768417250 Chr.16: 55846630-55846630 on Build GRCh38
rs16955807 Chr.16: 55846598-55846598 on Build GRCh38
rs771274526 Chr.16: 55846614-55846614 on Build GRCh38
rs11860488 Chr.16: 55846622-55846622 on Build GRCh38
rs780292496 Chr.16: 55846492-55846492 on Build GRCh38
rs370527653 Chr.16: 55846481-55846481 on Build GRCh38
rs544237697 Chr.16: 55846609-55846609 on Build GRCh38
rs778717480 Chr.16: 55846484-55846487 on Build GRCh38
rs750154282 Chr.16: 55846518-55846519 on Build GRCh38
rs747496710 Chr.16: 55846603-55846603 on Build GRCh38
rs142487845 Chr.16: 55846578-55846578 on Build GRCh38
rs773771399 Chr.16: 55846527-55846527 on Build GRCh38
rs752945026 Chr.16: 55846471-55846471 on Build GRCh38

View Full Product Details