Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49188181-49188442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf128
Entrez Gene ID
170371
Gene Name
chromosome 10 open reading frame 128
Gene Aliases
-
UniGene
Hs.385493
Gene Chromosome Location
Chr.10: 49154724-49188580 on Build GRCh38
Transcripts
NM_001010863.2 NM_001288740.1 NM_001288741.1 NM_001288742.1 NM_001288743.1 XM_011539378.2 XM_011539379.1 XM_011539380.1 XM_011539382.1 XM_011539383.2 XM_011539384.1 XM_011539385.1 XM_017015792.1 XM_017015793.1 XM_017015794.1 XM_017015795.1 XM_017015796.1 XM_017015797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3438297 c.12C>T p.F4F Chr.10: 49188301-49188301 on Build GRCh38
2145108 c.3G>A p.M1I Chr.10: 49188334-49188334 on Build GRCh38
3438296 c.36C>T p.F12F Chr.10: 49188301-49188301 on Build GRCh38
3867317 c.3G>A p.R1R Chr.10: 49188310-49188310 on Build GRCh38
6588090 c.30C>T p.I10I Chr.10: 49188307-49188307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746759621 Chr.10: 49188374-49188374 on Build GRCh38
rs576542301 Chr.10: 49188326-49188326 on Build GRCh38
rs753427117 Chr.10: 49188246-49188246 on Build GRCh38
rs776445358 Chr.10: 49188300-49188300 on Build GRCh38
rs759835709 Chr.10: 49188334-49188334 on Build GRCh38
rs141593519 Chr.10: 49188352-49188352 on Build GRCh38
rs754030549 Chr.10: 49188355-49188355 on Build GRCh38
rs779025187 Chr.10: 49188366-49188366 on Build GRCh38
rs751984505 Chr.10: 49188259-49188259 on Build GRCh38
rs76486037 Chr.10: 49188274-49188274 on Build GRCh38
rs769403820 Chr.10: 49188385-49188385 on Build GRCh38
rs1031555198 Chr.10: 49188368-49188368 on Build GRCh38
rs749317797 Chr.10: 49188264-49188264 on Build GRCh38
rs937320669 Chr.10: 49188410-49188410 on Build GRCh38
rs772984405 Chr.10: 49188305-49188305 on Build GRCh38
rs761430230 Chr.10: 49188243-49188243 on Build GRCh38
rs374627950 Chr.10: 49188301-49188301 on Build GRCh38
rs546449007 Chr.10: 49188278-49188278 on Build GRCh38
rs897698741 Chr.10: 49188309-49188309 on Build GRCh38
rs911476028 Chr.10: 49188256-49188256 on Build GRCh38
rs926054658 Chr.10: 49188388-49188388 on Build GRCh38
rs375419535 Chr.10: 49188223-49188223 on Build GRCh38
rs1014199419 Chr.10: 49188302-49188302 on Build GRCh38
rs768005021 Chr.10: 49188340-49188340 on Build GRCh38
rs376953936 Chr.10: 49188342-49188342 on Build GRCh38
rs781268972 Chr.10: 49188380-49188380 on Build GRCh38
rs766821150 Chr.10: 49188254-49188254 on Build GRCh38
rs966001095 Chr.10: 49188365-49188365 on Build GRCh38
rs937351677 Chr.10: 49188294-49188294 on Build GRCh38
rs375857671 Chr.10: 49188279-49188279 on Build GRCh38
rs764924403 Chr.10: 49188244-49188244 on Build GRCh38
rs994549347 Chr.10: 49188339-49188339 on Build GRCh38
rs61739968 Chr.10: 49188379-49188379 on Build GRCh38
rs779940052 Chr.10: 49188282-49188282 on Build GRCh38
rs573402818 Chr.10: 49188230-49188230 on Build GRCh38
rs766618129 Chr.10: 49188328-49188328 on Build GRCh38
rs17009534 Chr.10: 49188343-49188343 on Build GRCh38
rs184346486 Chr.10: 49188264-49188264 on Build GRCh38
rs529395359 Chr.10: 49188358-49188358 on Build GRCh38
rs1014017458 Chr.10: 49188401-49188401 on Build GRCh38
rs368989060 Chr.10: 49188298-49188298 on Build GRCh38
rs766723331 Chr.10: 49188332-49188332 on Build GRCh38
rs181582153 Chr.10: 49188263-49188263 on Build GRCh38
rs1000274456 Chr.10: 49188354-49188354 on Build GRCh38
rs768925960 Chr.10: 49188352-49188353 on Build GRCh38
rs371881496 Chr.10: 49188248-49188248 on Build GRCh38
rs746811750 Chr.10: 49188292-49188292 on Build GRCh38
rs1007655464 Chr.10: 49188349-49188349 on Build GRCh38
rs202214848 Chr.10: 49188265-49188265 on Build GRCh38
rs763403733 Chr.10: 49188250-49188250 on Build GRCh38
rs769493933 Chr.10: 49188304-49188304 on Build GRCh38
rs748151219 Chr.10: 49188381-49188381 on Build GRCh38

View Full Product Details