Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102047362-102047623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf70
Entrez Gene ID
85016
Gene Name
chromosome 11 open reading frame 70
Gene Aliases
-
UniGene
Hs.98328
Gene Chromosome Location
Chr.11: 102047409-102084560 on Build GRCh38
Transcripts
NM_001195005.1 NM_032930.2 XM_005271713.3 XM_017018454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317678 c.75G>C p.L25L Chr.11: 102047545-102047545 on Build GRCh38
4614105 c.23_24insCTTGGG p.G10_G11insLG Chr.11: 102047493-102047494 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs865809219 Chr.11: 102047581-102047581 on Build GRCh38
rs369244170 Chr.11: 102047397-102047397 on Build GRCh38
rs768247060 Chr.11: 102047586-102047586 on Build GRCh38
rs764496032 Chr.11: 102047505-102047505 on Build GRCh38
rs144077909 Chr.11: 102047409-102047409 on Build GRCh38
rs954099590 Chr.11: 102047572-102047572 on Build GRCh38
rs377307705 Chr.11: 102047414-102047414 on Build GRCh38
rs1046477484 Chr.11: 102047485-102047485 on Build GRCh38
rs986088466 Chr.11: 102047427-102047427 on Build GRCh38
rs1021009610 Chr.11: 102047392-102047392 on Build GRCh38
rs753634626 Chr.11: 102047493-102047494 on Build GRCh38
rs1005821766 Chr.11: 102047531-102047531 on Build GRCh38
rs918913130 Chr.11: 102047424-102047424 on Build GRCh38
rs966773844 Chr.11: 102047547-102047547 on Build GRCh38
rs748504645 Chr.11: 102047449-102047449 on Build GRCh38
rs1017426318 Chr.11: 102047599-102047599 on Build GRCh38
rs954517371 Chr.11: 102047406-102047406 on Build GRCh38
rs781589555 Chr.11: 102047441-102047441 on Build GRCh38
rs1007778077 Chr.11: 102047598-102047598 on Build GRCh38
rs917508727 Chr.11: 102047463-102047463 on Build GRCh38
rs1015882896 Chr.11: 102047561-102047561 on Build GRCh38
rs891796732 Chr.11: 102047527-102047527 on Build GRCh38
rs12804542 Chr.11: 102047596-102047596 on Build GRCh38
rs1055410333 Chr.11: 102047510-102047510 on Build GRCh38
rs879460198 Chr.11: 102047601-102047601 on Build GRCh38
rs948850801 Chr.11: 102047470-102047470 on Build GRCh38
rs756947261 Chr.11: 102047429-102047429 on Build GRCh38
rs1037430622 Chr.11: 102047440-102047440 on Build GRCh38
rs1030218172 Chr.11: 102047589-102047589 on Build GRCh38
rs529141845 Chr.11: 102047558-102047558 on Build GRCh38
rs770320127 Chr.11: 102047462-102047462 on Build GRCh38
rs868380721 Chr.11: 102047489-102047489 on Build GRCh38
rs1020957181 Chr.11: 102047533-102047533 on Build GRCh38
rs184947491 Chr.11: 102047486-102047486 on Build GRCh38
rs771478775 Chr.11: 102047515-102047515 on Build GRCh38
rs571717049 Chr.11: 102047396-102047396 on Build GRCh38
rs774795579 Chr.11: 102047520-102047520 on Build GRCh38
rs757196964 Chr.11: 102047591-102047632 on Build GRCh38
rs1030058401 Chr.11: 102047568-102047568 on Build GRCh38
rs114055321 Chr.11: 102047477-102047477 on Build GRCh38
rs941855020 Chr.11: 102047431-102047431 on Build GRCh38
rs897579291 Chr.11: 102047464-102047464 on Build GRCh38
rs1002853962 Chr.11: 102047506-102047506 on Build GRCh38

View Full Product Details