Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33301214-33301469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPA
Entrez Gene ID
1050
Gene Name
CCAAT/enhancer binding protein alpha
Gene Aliases
C/EBP-alpha, CEBP
UniGene
Hs.76171
Gene Chromosome Location
Chr.19: 33299934-33302564 on Build GRCh38
Transcripts
NM_001285829.1 NM_001287424.1 NM_001287435.1 NM_004364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
18411 c.990_991ins57 p.Q330_L331ins19 Chr.19: 33301424-33301425 on Build GRCh38
4169772 c.1055delA p.K352fs*>7 Chr.19: 33301360-33301360 on Build GRCh38
18478 c.1018G>A p.G340S Chr.19: 33301397-33301397 on Build GRCh38
4847185 c.1015C>T p.R339W Chr.19: 33301400-33301400 on Build GRCh38
5879629 c.953_954ins24 p.T318_S319insQQKVLELT Chr.19: 33301461-33301462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017549520 Chr.19: 33301284-33301284 on Build GRCh38
rs371719077 Chr.19: 33301320-33301320 on Build GRCh38
rs763641832 Chr.19: 33301298-33301298 on Build GRCh38
rs761752002 Chr.19: 33301381-33301381 on Build GRCh38
rs781549846 Chr.19: 33301442-33301442 on Build GRCh38
rs141430731 Chr.19: 33301374-33301374 on Build GRCh38
rs991316142 Chr.19: 33301252-33301252 on Build GRCh38
rs587778192 Chr.19: 33301406-33301406 on Build GRCh38
rs753024389 Chr.19: 33301323-33301323 on Build GRCh38
rs887167837 Chr.19: 33301246-33301246 on Build GRCh38
rs746226987 Chr.19: 33301350-33301350 on Build GRCh38
rs760372994 Chr.19: 33301405-33301405 on Build GRCh38
rs962903420 Chr.19: 33301288-33301288 on Build GRCh38
rs750194212 Chr.19: 33301310-33301310 on Build GRCh38
rs971289018 Chr.19: 33301275-33301275 on Build GRCh38
rs762447023 Chr.19: 33301290-33301290 on Build GRCh38
rs369632687 Chr.19: 33301415-33301415 on Build GRCh38
rs977188263 Chr.19: 33301325-33301325 on Build GRCh38
rs778040955 Chr.19: 33301330-33301330 on Build GRCh38
rs1051112472 Chr.19: 33301443-33301443 on Build GRCh38
rs775099224 Chr.19: 33301360-33301360 on Build GRCh38
rs757720364 Chr.19: 33301439-33301439 on Build GRCh38
rs543441756 Chr.19: 33301349-33301349 on Build GRCh38
rs774915318 Chr.19: 33301289-33301289 on Build GRCh38
rs375876732 Chr.19: 33301425-33301425 on Build GRCh38
rs868847139 Chr.19: 33301331-33301331 on Build GRCh38
rs758726582 Chr.19: 33301418-33301418 on Build GRCh38
rs375735989 Chr.19: 33301333-33301333 on Build GRCh38
rs773869623 Chr.19: 33301375-33301375 on Build GRCh38
rs543081565 Chr.19: 33301438-33301438 on Build GRCh38
rs148740083 Chr.19: 33301318-33301318 on Build GRCh38
rs767166906 Chr.19: 33301301-33301301 on Build GRCh38
rs376092627 Chr.19: 33301269-33301269 on Build GRCh38
rs758531608 Chr.19: 33301327-33301327 on Build GRCh38
rs373210526 Chr.19: 33301364-33301364 on Build GRCh38
rs372931505 Chr.19: 33301394-33301394 on Build GRCh38
rs771363863 Chr.19: 33301336-33301336 on Build GRCh38
rs770262748 Chr.19: 33301359-33301359 on Build GRCh38
rs577659401 Chr.19: 33301342-33301342 on Build GRCh38
rs375833261 Chr.19: 33301397-33301397 on Build GRCh38
rs748977615 Chr.19: 33301371-33301371 on Build GRCh38
rs767293321 Chr.19: 33301302-33301302 on Build GRCh38

View Full Product Details