Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150297928-150298185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSI
Entrez Gene ID
340075
Gene Name
arylsulfatase family member I
Gene Aliases
SPG66
UniGene
Hs.591252
Gene Chromosome Location
Chr.5: 150296346-150302962 on Build GRCh38
Transcripts
NM_001012301.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5417896 c.922A>G p.N308D Chr.5: 150298002-150298002 on Build GRCh38
7204076 c.749A>G p.Y250C Chr.5: 150298175-150298175 on Build GRCh38
5433928 c.800T>A p.V267E Chr.5: 150298124-150298124 on Build GRCh38
1064327 c.945G>T p.K315N Chr.5: 150297979-150297979 on Build GRCh38
206009 c.848G>A p.R283H Chr.5: 150298076-150298076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200086018 Chr.5: 150298077-150298077 on Build GRCh38
rs753465943 Chr.5: 150298094-150298094 on Build GRCh38
rs375582010 Chr.5: 150298062-150298062 on Build GRCh38
rs775948256 Chr.5: 150297970-150297970 on Build GRCh38
rs772303876 Chr.5: 150297967-150297967 on Build GRCh38
rs749936712 Chr.5: 150298102-150298102 on Build GRCh38
rs761975590 Chr.5: 150297989-150297989 on Build GRCh38
rs966027588 Chr.5: 150298029-150298029 on Build GRCh38
rs77878118 Chr.5: 150298030-150298030 on Build GRCh38
rs201541308 Chr.5: 150298096-150298096 on Build GRCh38
rs766051427 Chr.5: 150298101-150298101 on Build GRCh38
rs746414803 Chr.5: 150297964-150297964 on Build GRCh38
rs147161436 Chr.5: 150297991-150297991 on Build GRCh38
rs779388905 Chr.5: 150297961-150297961 on Build GRCh38
rs772663595 Chr.5: 150298019-150298019 on Build GRCh38
rs533440544 Chr.5: 150298145-150298145 on Build GRCh38
rs771369464 Chr.5: 150297960-150297960 on Build GRCh38
rs776993409 Chr.5: 150298057-150298057 on Build GRCh38
rs185169618 Chr.5: 150297983-150297983 on Build GRCh38
rs763826087 Chr.5: 150298146-150298146 on Build GRCh38
rs747762689 Chr.5: 150298006-150298007 on Build GRCh38
rs994655953 Chr.5: 150297987-150297987 on Build GRCh38
rs147391838 Chr.5: 150298051-150298051 on Build GRCh38
rs772864164 Chr.5: 150298041-150298043 on Build GRCh38
rs754848958 Chr.5: 150298133-150298133 on Build GRCh38
rs753554853 Chr.5: 150298004-150298004 on Build GRCh38
rs114744538 Chr.5: 150298123-150298123 on Build GRCh38
rs750097908 Chr.5: 150298157-150298157 on Build GRCh38
rs189890558 Chr.5: 150297990-150297990 on Build GRCh38
rs34775469 Chr.5: 150298148-150298149 on Build GRCh38
rs770975318 Chr.5: 150298142-150298142 on Build GRCh38
rs776520005 Chr.5: 150297984-150297984 on Build GRCh38
rs764879389 Chr.5: 150298154-150298154 on Build GRCh38
rs778824134 Chr.5: 150298166-150298166 on Build GRCh38
rs746683228 Chr.5: 150298056-150298058 on Build GRCh38
rs778442199 Chr.5: 150298006-150298006 on Build GRCh38
rs765851189 Chr.5: 150298164-150298164 on Build GRCh38
rs749401740 Chr.5: 150298140-150298140 on Build GRCh38
rs780214141 Chr.5: 150298025-150298025 on Build GRCh38
rs763835530 Chr.5: 150298090-150298090 on Build GRCh38
rs142802678 Chr.5: 150298012-150298012 on Build GRCh38
rs770174912 Chr.5: 150298134-150298134 on Build GRCh38
rs773450988 Chr.5: 150298064-150298064 on Build GRCh38
rs145206443 Chr.5: 150298112-150298112 on Build GRCh38
rs762169944 Chr.5: 150298061-150298061 on Build GRCh38
rs748800109 Chr.5: 150297958-150297958 on Build GRCh38
rs201644621 Chr.5: 150298135-150298135 on Build GRCh38
rs141695719 Chr.5: 150297954-150297954 on Build GRCh38
rs140546154 Chr.5: 150297994-150297994 on Build GRCh38
rs926772891 Chr.5: 150298100-150298100 on Build GRCh38
rs139203947 Chr.5: 150298072-150298072 on Build GRCh38
rs758770660 Chr.5: 150298105-150298105 on Build GRCh38
rs150250495 Chr.5: 150297995-150297995 on Build GRCh38
rs781725394 Chr.5: 150298132-150298132 on Build GRCh38
rs200155745 Chr.5: 150298003-150298003 on Build GRCh38
rs761385589 Chr.5: 150298152-150298152 on Build GRCh38
rs149948064 Chr.5: 150298076-150298076 on Build GRCh38
rs776440347 Chr.5: 150298148-150298148 on Build GRCh38
rs775494433 Chr.5: 150298143-150298143 on Build GRCh38
rs141392111 Chr.5: 150297957-150297957 on Build GRCh38
rs771488404 Chr.5: 150298016-150298016 on Build GRCh38
rs757916929 Chr.5: 150298161-150298161 on Build GRCh38
rs147972506 Chr.5: 150297953-150297953 on Build GRCh38

View Full Product Details