Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 146116788-146117061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
UniGene
Hs.655684
Gene Chromosome Location
Chr.7: 146116035-148420998 on Build GRCh38
Transcripts
NM_014141.5 XM_017011950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669704 c.73G>A p.A25T Chr.7: 146116949-146116949 on Build GRCh38
5558674 c.44T>G p.L15R Chr.7: 146116920-146116920 on Build GRCh38
4961903 c.9G>A p.A3A Chr.7: 146116885-146116885 on Build GRCh38
6287579 c.20C>A p.A7D Chr.7: 146116896-146116896 on Build GRCh38
5345284 c.18C>T p.R6R Chr.7: 146116894-146116894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978368135 Chr.7: 146116958-146116958 on Build GRCh38
rs886062048 Chr.7: 146116935-146116935 on Build GRCh38
rs762828266 Chr.7: 146116841-146116841 on Build GRCh38
rs572889297 Chr.7: 146116966-146116966 on Build GRCh38
rs982512594 Chr.7: 146116925-146116925 on Build GRCh38
rs796052375 Chr.7: 146116913-146116913 on Build GRCh38
rs886062047 Chr.7: 146116816-146116816 on Build GRCh38
rs376828245 Chr.7: 146116829-146116829 on Build GRCh38
rs1029991366 Chr.7: 146116851-146116851 on Build GRCh38
rs759610282 Chr.7: 146117014-146117014 on Build GRCh38
rs139443887 Chr.7: 146117022-146117022 on Build GRCh38
rs1005196299 Chr.7: 146116825-146116825 on Build GRCh38
rs1050674953 Chr.7: 146117020-146117020 on Build GRCh38
rs200866893 Chr.7: 146116949-146116949 on Build GRCh38
rs748908765 Chr.7: 146116894-146116894 on Build GRCh38
rs748064330 Chr.7: 146116971-146116971 on Build GRCh38
rs587780903 Chr.7: 146116845-146116845 on Build GRCh38
rs939547083 Chr.7: 146117006-146117006 on Build GRCh38
rs796052366 Chr.7: 146116959-146116959 on Build GRCh38
rs886062049 Chr.7: 146116963-146116963 on Build GRCh38
rs780831072 Chr.7: 146116848-146116848 on Build GRCh38
rs542709358 Chr.7: 146116916-146116916 on Build GRCh38
rs796052368 Chr.7: 146116831-146116831 on Build GRCh38
rs762640925 Chr.7: 146116974-146116975 on Build GRCh38
rs1017715989 Chr.7: 146116915-146116915 on Build GRCh38
rs942423011 Chr.7: 146117033-146117033 on Build GRCh38
rs794726938 Chr.7: 146116973-146116973 on Build GRCh38
rs879368718 Chr.7: 146116859-146116859 on Build GRCh38
rs549396215 Chr.7: 146116828-146116828 on Build GRCh38
rs768374052 Chr.7: 146116941-146116941 on Build GRCh38
rs755613342 Chr.7: 146116852-146116852 on Build GRCh38
rs375492857 Chr.7: 146116995-146116995 on Build GRCh38
rs886044073 Chr.7: 146116924-146116924 on Build GRCh38
rs191830389 Chr.7: 146116842-146116842 on Build GRCh38
rs538552704 Chr.7: 146116823-146116823 on Build GRCh38
rs886044541 Chr.7: 146116905-146116905 on Build GRCh38
rs775422890 Chr.7: 146116835-146116835 on Build GRCh38
rs773052825 Chr.7: 146117000-146117000 on Build GRCh38
rs571236387 Chr.7: 146116808-146116808 on Build GRCh38
rs779710846 Chr.7: 146116890-146116890 on Build GRCh38
rs972116002 Chr.7: 146116974-146116974 on Build GRCh38

View Full Product Details