Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32734891-32735140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030239 c.2677C>T p.R893* Chr.21: 32735027-32735027 on Build GRCh38
7099118 c.2689C>T p.H897Y Chr.21: 32735015-32735015 on Build GRCh38
257010 c.2660A>C p.K887T Chr.21: 32735044-32735044 on Build GRCh38
1030238 c.2689C>A p.H897N Chr.21: 32735015-32735015 on Build GRCh38
6489374 c.2694T>A p.A898A Chr.21: 32735010-32735010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770144337 Chr.21: 32734987-32734987 on Build GRCh38
rs977439581 Chr.21: 32734940-32734940 on Build GRCh38
rs746260675 Chr.21: 32734948-32734948 on Build GRCh38
rs752157441 Chr.21: 32734924-32734924 on Build GRCh38
rs917535419 Chr.21: 32734932-32734932 on Build GRCh38
rs780014199 Chr.21: 32735100-32735101 on Build GRCh38
rs116339270 Chr.21: 32734962-32734962 on Build GRCh38
rs780259389 Chr.21: 32735112-32735112 on Build GRCh38
rs940086731 Chr.21: 32735022-32735022 on Build GRCh38
rs752210605 Chr.21: 32735082-32735082 on Build GRCh38
rs755797875 Chr.21: 32734933-32734933 on Build GRCh38
rs773248636 Chr.21: 32734992-32734992 on Build GRCh38
rs201458089 Chr.21: 32735084-32735084 on Build GRCh38
rs766080046 Chr.21: 32735008-32735008 on Build GRCh38
rs768905578 Chr.21: 32734968-32734968 on Build GRCh38
rs374822523 Chr.21: 32735106-32735106 on Build GRCh38
rs376469469 Chr.21: 32735089-32735089 on Build GRCh38
rs774382781 Chr.21: 32735032-32735032 on Build GRCh38
rs762861716 Chr.21: 32734996-32734996 on Build GRCh38
rs753429701 Chr.21: 32735092-32735092 on Build GRCh38
rs905693171 Chr.21: 32735091-32735091 on Build GRCh38
rs935990060 Chr.21: 32734994-32734994 on Build GRCh38
rs372725852 Chr.21: 32734945-32734945 on Build GRCh38
rs776872837 Chr.21: 32734976-32734976 on Build GRCh38
rs200465600 Chr.21: 32734923-32734923 on Build GRCh38
rs750885622 Chr.21: 32734927-32734928 on Build GRCh38
rs759496681 Chr.21: 32735034-32735034 on Build GRCh38
rs200811797 Chr.21: 32735001-32735001 on Build GRCh38
rs1039801475 Chr.21: 32735048-32735048 on Build GRCh38
rs143512881 Chr.21: 32734961-32734961 on Build GRCh38
rs972702748 Chr.21: 32735026-32735026 on Build GRCh38
rs376031602 Chr.21: 32734967-32734967 on Build GRCh38
rs547478950 Chr.21: 32734982-32734982 on Build GRCh38
rs767406461 Chr.21: 32735064-32735064 on Build GRCh38
rs899908330 Chr.21: 32735052-32735052 on Build GRCh38
rs756581949 Chr.21: 32734987-32734988 on Build GRCh38
rs748197981 Chr.21: 32734986-32734986 on Build GRCh38

View Full Product Details