Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70329482-70329707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
UniGene
Hs.133350
Gene Chromosome Location
Chr.12: 70242993-70354993 on Build GRCh38
Transcripts
NM_001199302.1 NM_001199303.1 NM_014515.5 NR_037615.1 XM_006719429.3 XM_006719430.3 XM_006719432.3 XM_006719433.3 XM_011538399.1 XM_011538400.2 XM_017019348.1 XM_017019349.1 XM_017019350.1 XM_017019351.1 XM_017019352.1 XM_017019353.1 XM_017019354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
943103 c.328G>A p.V110I Chr.12: 70329512-70329512 on Build GRCh38
6631448 c.333G>A p.T111T Chr.12: 70329517-70329517 on Build GRCh38
943102 c.327C>T p.H109H Chr.12: 70329511-70329511 on Build GRCh38
1316403 c.371C>G p.P124R Chr.12: 70329555-70329555 on Build GRCh38
1316402 c.370C>T p.P124S Chr.12: 70329554-70329554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779061066 Chr.12: 70329620-70329620 on Build GRCh38
rs919265194 Chr.12: 70329676-70329687 on Build GRCh38
rs181208252 Chr.12: 70329505-70329505 on Build GRCh38
rs766711761 Chr.12: 70329584-70329584 on Build GRCh38
rs756710697 Chr.12: 70329616-70329616 on Build GRCh38
rs750491908 Chr.12: 70329529-70329529 on Build GRCh38
rs377546399 Chr.12: 70329517-70329517 on Build GRCh38
rs927062896 Chr.12: 70329550-70329550 on Build GRCh38
rs992577525 Chr.12: 70329674-70329674 on Build GRCh38
rs765785036 Chr.12: 70329532-70329532 on Build GRCh38
rs761729984 Chr.12: 70329530-70329530 on Build GRCh38
rs1021501343 Chr.12: 70329603-70329603 on Build GRCh38
rs755824919 Chr.12: 70329602-70329602 on Build GRCh38
rs758317551 Chr.12: 70329585-70329585 on Build GRCh38
rs371050057 Chr.12: 70329607-70329607 on Build GRCh38
rs923938669 Chr.12: 70329643-70329643 on Build GRCh38
rs980876424 Chr.12: 70329630-70329630 on Build GRCh38
rs926713135 Chr.12: 70329634-70329634 on Build GRCh38
rs960711795 Chr.12: 70329668-70329668 on Build GRCh38
rs750913052 Chr.12: 70329552-70329552 on Build GRCh38
rs779079258 Chr.12: 70329516-70329516 on Build GRCh38
rs915420738 Chr.12: 70329604-70329604 on Build GRCh38

View Full Product Details