Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116121373-116121598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL10A1
Entrez Gene ID
1300
Gene Name
collagen type X alpha 1 chain
Gene Aliases
-
UniGene
Hs.520339
Gene Chromosome Location
Chr.6: 116118909-116218698 on Build GRCh38
Transcripts
NM_000493.3 XM_006715333.3 XM_011535432.2 XM_011535433.2 XM_017010248.1 XM_017010249.1 NM_152729.2 XM_006715378.3

Gene Symbol
NT5DC1
Entrez Gene ID
221294
Gene Name
5'-nucleotidase domain containing 1
Gene Aliases
C6orf200, LP2642, NT5C2L1
UniGene
Hs.520341
Gene Chromosome Location
Chr.6: 116100833-116245690 on Build GRCh38
Transcripts
NM_000493.3 XM_006715333.3 XM_011535432.2 XM_011535433.2 XM_017010248.1 XM_017010249.1 NM_152729.2 XM_006715378.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3393774 c.628C>A p.P210T Chr.6: 116121488-116121488 on Build GRCh38
7361924 c.713G>T p.G238V Chr.6: 116121403-116121403 on Build GRCh38
5584850 c.710G>A p.R237K Chr.6: 116121406-116121406 on Build GRCh38
6254518 c.742C>A p.P248T Chr.6: 116121374-116121374 on Build GRCh38
319571 c.518C>A p.P173H Chr.6: 116121598-116121598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759656228 Chr.6: 116121449-116121449 on Build GRCh38
rs769103179 Chr.6: 116121524-116121524 on Build GRCh38
rs772549860 Chr.6: 116121451-116121451 on Build GRCh38
rs1023428679 Chr.6: 116121438-116121438 on Build GRCh38
rs578248225 Chr.6: 116121505-116121505 on Build GRCh38
rs771118636 Chr.6: 116121445-116121445 on Build GRCh38
rs779159900 Chr.6: 116121514-116121514 on Build GRCh38
rs754927069 Chr.6: 116121573-116121573 on Build GRCh38
rs202127284 Chr.6: 116121398-116121398 on Build GRCh38
rs148785195 Chr.6: 116121523-116121523 on Build GRCh38
rs780179251 Chr.6: 116121405-116121405 on Build GRCh38
rs775927537 Chr.6: 116121460-116121460 on Build GRCh38
rs761512500 Chr.6: 116121481-116121481 on Build GRCh38
rs907207713 Chr.6: 116121497-116121497 on Build GRCh38
rs772100888 Chr.6: 116121521-116121521 on Build GRCh38
rs921724547 Chr.6: 116121490-116121490 on Build GRCh38
rs202057046 Chr.6: 116121430-116121430 on Build GRCh38
rs780876635 Chr.6: 116121413-116121413 on Build GRCh38
rs767997583 Chr.6: 116121457-116121458 on Build GRCh38
rs764863859 Chr.6: 116121484-116121484 on Build GRCh38
rs754168768 Chr.6: 116121472-116121472 on Build GRCh38
rs764919633 Chr.6: 116121544-116121544 on Build GRCh38
rs754873544 Chr.6: 116121508-116121508 on Build GRCh38
rs1053242460 Chr.6: 116121526-116121526 on Build GRCh38
rs777047241 Chr.6: 116121530-116121530 on Build GRCh38
rs779728797 Chr.6: 116121493-116121493 on Build GRCh38
rs1021780104 Chr.6: 116121571-116121571 on Build GRCh38
rs751499436 Chr.6: 116121504-116121504 on Build GRCh38
rs762721417 Chr.6: 116121551-116121551 on Build GRCh38
rs142492015 Chr.6: 116121446-116121446 on Build GRCh38
rs560652096 Chr.6: 116121541-116121541 on Build GRCh38
rs544895054 Chr.6: 116121488-116121488 on Build GRCh38
rs747962638 Chr.6: 116121513-116121513 on Build GRCh38
rs760913730 Chr.6: 116121505-116121506 on Build GRCh38
rs1027287541 Chr.6: 116121392-116121392 on Build GRCh38
rs950122683 Chr.6: 116121566-116121566 on Build GRCh38
rs751125454 Chr.6: 116121561-116121561 on Build GRCh38
rs746116127 Chr.6: 116121519-116121519 on Build GRCh38
rs768444506 Chr.6: 116121408-116121408 on Build GRCh38
rs749457223 Chr.6: 116121434-116121434 on Build GRCh38
rs201737018 Chr.6: 116121549-116121549 on Build GRCh38
rs112875740 Chr.6: 116121396-116121396 on Build GRCh38
rs746777649 Chr.6: 116121407-116121407 on Build GRCh38
rs765917029 Chr.6: 116121555-116121555 on Build GRCh38
rs146114911 Chr.6: 116121491-116121491 on Build GRCh38
rs886060992 Chr.6: 116121483-116121483 on Build GRCh38
rs761203337 Chr.6: 116121463-116121463 on Build GRCh38
rs369551991 Chr.6: 116121395-116121395 on Build GRCh38
rs773630614 Chr.6: 116121490-116121490 on Build GRCh38
rs575467695 Chr.6: 116121498-116121498 on Build GRCh38

View Full Product Details