Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31388826-31389097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB119
Entrez Gene ID
245932
Gene Name
defensin beta 119
Gene Aliases
DEFB-19, DEFB-20, DEFB120, DEFB20, ESC42-RELA, ESC42-RELB
UniGene
Hs.516998
Gene Chromosome Location
Chr.20: 31377163-31390649 on Build GRCh38
Transcripts
NM_001271209.1 NM_153289.3 NM_153323.4 NR_073151.1 NR_073152.1 NR_073153.1 NR_126440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
576837 c.243G>A p.M81I Chr.20: 31389049-31389049 on Build GRCh38
2888359 c.267G>A p.*89* Chr.20: 31389025-31389025 on Build GRCh38
1025542 c.267G>T p.*89Y Chr.20: 31389025-31389025 on Build GRCh38
1025543 c.238A>C p.K80Q Chr.20: 31389054-31389054 on Build GRCh38
4399043 c.203G>A p.G68D Chr.20: 31389089-31389089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111410523 Chr.20: 31388953-31388953 on Build GRCh38
rs749194522 Chr.20: 31389050-31389050 on Build GRCh38
rs868690364 Chr.20: 31389035-31389035 on Build GRCh38
rs866493529 Chr.20: 31388977-31388977 on Build GRCh38
rs755301489 Chr.20: 31388974-31388974 on Build GRCh38
rs12329612 Chr.20: 31389027-31389027 on Build GRCh38
rs1008376723 Chr.20: 31388973-31388973 on Build GRCh38
rs908863058 Chr.20: 31388873-31388873 on Build GRCh38
rs769628042 Chr.20: 31389020-31389020 on Build GRCh38
rs573553340 Chr.20: 31388963-31388963 on Build GRCh38
rs151051085 Chr.20: 31388916-31388916 on Build GRCh38
rs781542518 Chr.20: 31389014-31389014 on Build GRCh38
rs991295383 Chr.20: 31388884-31388884 on Build GRCh38
rs112691733 Chr.20: 31388879-31388879 on Build GRCh38
rs758419565 Chr.20: 31388998-31388998 on Build GRCh38
rs745699187 Chr.20: 31389019-31389019 on Build GRCh38
rs772844037 Chr.20: 31389075-31389075 on Build GRCh38
rs765728376 Chr.20: 31388976-31388976 on Build GRCh38
rs768058751 Chr.20: 31389060-31389060 on Build GRCh38
rs571302061 Chr.20: 31388939-31388939 on Build GRCh38
rs372022906 Chr.20: 31388999-31388999 on Build GRCh38
rs753538807 Chr.20: 31388854-31388854 on Build GRCh38
rs73108004 Chr.20: 31388954-31388954 on Build GRCh38
rs902851368 Chr.20: 31388930-31388930 on Build GRCh38
rs151140528 Chr.20: 31389067-31389067 on Build GRCh38
rs938495512 Chr.20: 31388892-31388892 on Build GRCh38
rs761285484 Chr.20: 31389074-31389074 on Build GRCh38
rs764995997 Chr.20: 31388866-31388866 on Build GRCh38
rs757504624 Chr.20: 31389010-31389010 on Build GRCh38
rs557521084 Chr.20: 31388918-31388918 on Build GRCh38
rs754737390 Chr.20: 31388855-31388855 on Build GRCh38
rs6057686 Chr.20: 31388904-31388904 on Build GRCh38
rs753231988 Chr.20: 31388997-31388997 on Build GRCh38
rs947271954 Chr.20: 31388908-31388908 on Build GRCh38
rs189306146 Chr.20: 31388890-31388890 on Build GRCh38
rs890182711 Chr.20: 31388964-31388964 on Build GRCh38
rs915853273 Chr.20: 31388903-31388903 on Build GRCh38
rs866508552 Chr.20: 31389049-31389049 on Build GRCh38
rs928461889 Chr.20: 31388891-31388891 on Build GRCh38

View Full Product Details