Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26156351-26156618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H1E
Entrez Gene ID
3008
Gene Name
histone cluster 1, H1e
Gene Aliases
H1.4, H1E, H1F4, H1s-4, dJ221C16.5
UniGene
Hs.248133
Gene Chromosome Location
Chr.6: 26156331-26157115 on Build GRCh38
Transcripts
NM_005321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884345 c.184C>A p.L62I Chr.6: 26156574-26156574 on Build GRCh38
1292259 c.214_219delGACGTG p.D72_V73delDV Chr.6: 26156604-26156609 on Build GRCh38
7428664 c.226_228delAAC p.N77delN Chr.6: 26156616-26156618 on Build GRCh38
4832277 c.153C>T p.S51S Chr.6: 26156543-26156543 on Build GRCh38
5948934 c.127C>G p.L43V Chr.6: 26156517-26156517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759683845 Chr.6: 26156490-26156490 on Build GRCh38
rs762852197 Chr.6: 26156460-26156460 on Build GRCh38
rs771008119 Chr.6: 26156506-26156506 on Build GRCh38
rs777296172 Chr.6: 26156379-26156379 on Build GRCh38
rs1047740849 Chr.6: 26156550-26156550 on Build GRCh38
rs754791514 Chr.6: 26156373-26156373 on Build GRCh38
rs765638416 Chr.6: 26156491-26156491 on Build GRCh38
rs1017769394 Chr.6: 26156560-26156560 on Build GRCh38
rs377472965 Chr.6: 26156539-26156539 on Build GRCh38
rs138821617 Chr.6: 26156524-26156524 on Build GRCh38
rs760029136 Chr.6: 26156569-26156569 on Build GRCh38
rs767992734 Chr.6: 26156545-26156545 on Build GRCh38
rs775335777 Chr.6: 26156443-26156443 on Build GRCh38
rs373759542 Chr.6: 26156535-26156535 on Build GRCh38
rs760978374 Chr.6: 26156582-26156582 on Build GRCh38
rs1030953348 Chr.6: 26156422-26156422 on Build GRCh38
rs780842942 Chr.6: 26156456-26156456 on Build GRCh38
rs771979983 Chr.6: 26156503-26156503 on Build GRCh38
rs200756277 Chr.6: 26156576-26156576 on Build GRCh38
rs759953648 Chr.6: 26156382-26156382 on Build GRCh38
rs141942142 Chr.6: 26156530-26156530 on Build GRCh38
rs747063072 Chr.6: 26156377-26156377 on Build GRCh38
rs772890562 Chr.6: 26156429-26156429 on Build GRCh38
rs763754773 Chr.6: 26156585-26156585 on Build GRCh38
rs755659578 Chr.6: 26156497-26156497 on Build GRCh38
rs766638957 Chr.6: 26156484-26156484 on Build GRCh38
rs774751617 Chr.6: 26156372-26156372 on Build GRCh38
rs757147711 Chr.6: 26156593-26156593 on Build GRCh38
rs745963346 Chr.6: 26156424-26156424 on Build GRCh38
rs759410762 Chr.6: 26156416-26156416 on Build GRCh38
rs761734336 Chr.6: 26156464-26156464 on Build GRCh38
rs762441962 Chr.6: 26156574-26156574 on Build GRCh38
rs201669716 Chr.6: 26156558-26156558 on Build GRCh38
rs765930149 Chr.6: 26156517-26156517 on Build GRCh38
rs367916158 Chr.6: 26156387-26156387 on Build GRCh38
rs374780607 Chr.6: 26156392-26156392 on Build GRCh38
rs772034993 Chr.6: 26156482-26156482 on Build GRCh38
rs139757464 Chr.6: 26156561-26156561 on Build GRCh38
rs755384846 Chr.6: 26156445-26156445 on Build GRCh38
rs567153678 Chr.6: 26156505-26156505 on Build GRCh38
rs535188796 Chr.6: 26156579-26156579 on Build GRCh38
rs776052237 Chr.6: 26156439-26156439 on Build GRCh38
rs148757707 Chr.6: 26156594-26156594 on Build GRCh38
rs754736485 Chr.6: 26156390-26156390 on Build GRCh38
rs745358897 Chr.6: 26156457-26156457 on Build GRCh38
rs770963016 Chr.6: 26156404-26156404 on Build GRCh38
rs980627455 Chr.6: 26156531-26156531 on Build GRCh38
rs369107797 Chr.6: 26156475-26156475 on Build GRCh38
rs752502613 Chr.6: 26156396-26156396 on Build GRCh38
rs370379784 Chr.6: 26156501-26156501 on Build GRCh38
rs752204746 Chr.6: 26156556-26156556 on Build GRCh38
rs779505837 Chr.6: 26156474-26156474 on Build GRCh38
rs781197204 Chr.6: 26156418-26156418 on Build GRCh38
rs368825996 Chr.6: 26156374-26156374 on Build GRCh38
rs986521398 Chr.6: 26156434-26156434 on Build GRCh38
rs35578662 Chr.6: 26156507-26156507 on Build GRCh38
rs150634482 Chr.6: 26156513-26156513 on Build GRCh38
rs759075864 Chr.6: 26156441-26156441 on Build GRCh38
rs954509244 Chr.6: 26156518-26156518 on Build GRCh38
rs769960678 Chr.6: 26156565-26156565 on Build GRCh38
rs771718049 Chr.6: 26156412-26156412 on Build GRCh38
rs376608867 Chr.6: 26156499-26156499 on Build GRCh38
rs569775913 Chr.6: 26156559-26156559 on Build GRCh38
rs777887295 Chr.6: 26156400-26156400 on Build GRCh38
rs377661226 Chr.6: 26156471-26156471 on Build GRCh38
rs374113105 Chr.6: 26156465-26156465 on Build GRCh38
rs755955332 Chr.6: 26156596-26156596 on Build GRCh38
rs781753197 Chr.6: 26156405-26156405 on Build GRCh38
rs770273263 Chr.6: 26156409-26156409 on Build GRCh38
rs374200136 Chr.6: 26156567-26156567 on Build GRCh38
rs373802381 Chr.6: 26156478-26156478 on Build GRCh38
rs776546273 Chr.6: 26156415-26156415 on Build GRCh38
rs757889192 Chr.6: 26156459-26156459 on Build GRCh38
rs2298089 Chr.6: 26156371-26156371 on Build GRCh38
rs757367774 Chr.6: 26156446-26156446 on Build GRCh38
rs769330993 Chr.6: 26156458-26156458 on Build GRCh38
rs767093143 Chr.6: 26156385-26156385 on Build GRCh38
rs541308925 Chr.6: 26156445-26156445 on Build GRCh38
rs764987875 Chr.6: 26156552-26156552 on Build GRCh38
rs547695315 Chr.6: 26156529-26156529 on Build GRCh38
rs368341452 Chr.6: 26156573-26156573 on Build GRCh38
rs780734214 Chr.6: 26156556-26156557 on Build GRCh38
rs144396504 Chr.6: 26156583-26156583 on Build GRCh38
rs374382666 Chr.6: 26156378-26156378 on Build GRCh38
rs761619834 Chr.6: 26156384-26156384 on Build GRCh38
rs748838528 Chr.6: 26156479-26156479 on Build GRCh38
rs963697864 Chr.6: 26156440-26156440 on Build GRCh38
rs773954612 Chr.6: 26156462-26156462 on Build GRCh38
rs762056816 Chr.6: 26156543-26156543 on Build GRCh38
rs753820303 Chr.6: 26156487-26156487 on Build GRCh38
rs568154277 Chr.6: 26156516-26156516 on Build GRCh38
rs772522986 Chr.6: 26156545-26156546 on Build GRCh38
rs146982083 Chr.6: 26156495-26156495 on Build GRCh38
rs200564842 Chr.6: 26156586-26156586 on Build GRCh38
rs768953669 Chr.6: 26156428-26156428 on Build GRCh38
rs751765449 Chr.6: 26156421-26156421 on Build GRCh38
rs145783632 Chr.6: 26156449-26156449 on Build GRCh38
rs551818027 Chr.6: 26156502-26156502 on Build GRCh38
rs200323601 Chr.6: 26156504-26156504 on Build GRCh38
rs199978177 Chr.6: 26156435-26156435 on Build GRCh38
rs371530012 Chr.6: 26156411-26156411 on Build GRCh38
rs969006148 Chr.6: 26156454-26156454 on Build GRCh38
rs778623700 Chr.6: 26156526-26156526 on Build GRCh38
rs754602998 Chr.6: 26156549-26156549 on Build GRCh38
rs146311252 Chr.6: 26156537-26156537 on Build GRCh38
rs762547996 Chr.6: 26156584-26156584 on Build GRCh38
rs769917587 Chr.6: 26156538-26156538 on Build GRCh38
rs748081668 Chr.6: 26156419-26156419 on Build GRCh38
rs753419578 Chr.6: 26156546-26156546 on Build GRCh38
rs753242501 Chr.6: 26156494-26156494 on Build GRCh38
rs756624705 Chr.6: 26156425-26156425 on Build GRCh38
rs774370544 Chr.6: 26156480-26156480 on Build GRCh38
rs768767088 Chr.6: 26156572-26156572 on Build GRCh38
rs765164128 Chr.6: 26156417-26156417 on Build GRCh38
rs138910780 Chr.6: 26156468-26156468 on Build GRCh38
rs780789434 Chr.6: 26156476-26156476 on Build GRCh38
rs776712433 Chr.6: 26156536-26156536 on Build GRCh38
rs545095988 Chr.6: 26156450-26156452 on Build GRCh38
rs141466409 Chr.6: 26156483-26156483 on Build GRCh38
rs756055232 Chr.6: 26156519-26156519 on Build GRCh38
rs760212562 Chr.6: 26156414-26156414 on Build GRCh38
rs919941742 Chr.6: 26156544-26156544 on Build GRCh38
rs749801188 Chr.6: 26156448-26156448 on Build GRCh38
rs765109173 Chr.6: 26156442-26156442 on Build GRCh38
rs748550045 Chr.6: 26156450-26156450 on Build GRCh38
rs982441541 Chr.6: 26156588-26156588 on Build GRCh38
rs755373320 Chr.6: 26156472-26156472 on Build GRCh38
rs766291763 Chr.6: 26156388-26156388 on Build GRCh38
rs201889498 Chr.6: 26156453-26156453 on Build GRCh38
rs779790843 Chr.6: 26156498-26156498 on Build GRCh38
rs774145855 Chr.6: 26156383-26156383 on Build GRCh38
rs144551358 Chr.6: 26156564-26156564 on Build GRCh38
rs750493363 Chr.6: 26156447-26156447 on Build GRCh38
rs371861388 Chr.6: 26156375-26156375 on Build GRCh38
rs951047896 Chr.6: 26156523-26156523 on Build GRCh38
rs766435782 Chr.6: 26156470-26156470 on Build GRCh38
rs747676333 Chr.6: 26156528-26156528 on Build GRCh38
rs369184031 Chr.6: 26156401-26156401 on Build GRCh38
rs555676703 Chr.6: 26156426-26156426 on Build GRCh38
rs371691151 Chr.6: 26156389-26156389 on Build GRCh38
rs376720047 Chr.6: 26156423-26156423 on Build GRCh38
rs764538039 Chr.6: 26156540-26156540 on Build GRCh38
rs761681415 Chr.6: 26156481-26156481 on Build GRCh38
rs748723810 Chr.6: 26156499-26156499 on Build GRCh38

View Full Product Details