Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64284097-64284330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAD
Entrez Gene ID
572
Gene Name
BCL2 associated agonist of cell death
Gene Aliases
BBC2, BCL2L8
UniGene
Hs.370254
Gene Chromosome Location
Chr.11: 64269828-64284704 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018016.1

Gene Symbol
GPR137
Entrez Gene ID
56834
Gene Name
G protein-coupled receptor 137
Gene Aliases
C11orf4, GPR137A, TM7SF1L1
UniGene
Hs.523763
Gene Chromosome Location
Chr.11: 64270337-64289500 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5721722 c.157G>A p.E53K Chr.11: 64284212-64284212 on Build GRCh38
3953615 c.128G>C p.G43A Chr.11: 64284241-64284241 on Build GRCh38
4656508 c.90G>A p.G30G Chr.11: 64284279-64284279 on Build GRCh38
7403730 c.85_90delGACGGG p.D29_G30delDG Chr.11: 64284279-64284284 on Build GRCh38
4825240 c.95C>G p.S32* Chr.11: 64284274-64284274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776845441 Chr.11: 64284171-64284171 on Build GRCh38
rs142604226 Chr.11: 64284216-64284216 on Build GRCh38
rs750070533 Chr.11: 64284139-64284139 on Build GRCh38
rs759558076 Chr.11: 64284180-64284180 on Build GRCh38
rs763357865 Chr.11: 64284298-64284298 on Build GRCh38
rs778657393 Chr.11: 64284153-64284153 on Build GRCh38
rs753463626 Chr.11: 64284149-64284149 on Build GRCh38
rs777661113 Chr.11: 64284164-64284164 on Build GRCh38
rs11541683 Chr.11: 64284227-64284227 on Build GRCh38
rs755881643 Chr.11: 64284141-64284141 on Build GRCh38
rs1039826529 Chr.11: 64284213-64284213 on Build GRCh38
rs199973522 Chr.11: 64284234-64284234 on Build GRCh38
rs752542699 Chr.11: 64284237-64284237 on Build GRCh38
rs201722570 Chr.11: 64284293-64284293 on Build GRCh38
rs367948500 Chr.11: 64284248-64284248 on Build GRCh38
rs147750998 Chr.11: 64284201-64284201 on Build GRCh38
rs769943605 Chr.11: 64284199-64284199 on Build GRCh38
rs569767797 Chr.11: 64284290-64284290 on Build GRCh38
rs766061149 Chr.11: 64284223-64284223 on Build GRCh38
rs372012989 Chr.11: 64284134-64284134 on Build GRCh38
rs764551221 Chr.11: 64284136-64284136 on Build GRCh38
rs372175324 Chr.11: 64284266-64284266 on Build GRCh38
rs762208923 Chr.11: 64284308-64284308 on Build GRCh38
rs769270284 Chr.11: 64284306-64284306 on Build GRCh38
rs748135213 Chr.11: 64284160-64284160 on Build GRCh38
rs763555901 Chr.11: 64284202-64284202 on Build GRCh38
rs376887057 Chr.11: 64284151-64284151 on Build GRCh38
rs770729654 Chr.11: 64284274-64284274 on Build GRCh38
rs775688969 Chr.11: 64284295-64284295 on Build GRCh38
rs764345127 Chr.11: 64284203-64284203 on Build GRCh38
rs771721252 Chr.11: 64284161-64284161 on Build GRCh38
rs754521057 Chr.11: 64284150-64284150 on Build GRCh38
rs758407783 Chr.11: 64284247-64284247 on Build GRCh38
rs777335964 Chr.11: 64284276-64284276 on Build GRCh38
rs753264880 Chr.11: 64284133-64284133 on Build GRCh38
rs367613306 Chr.11: 64284165-64284165 on Build GRCh38
rs771212031 Chr.11: 64284166-64284166 on Build GRCh38
rs752030848 Chr.11: 64284215-64284215 on Build GRCh38
rs151235996 Chr.11: 64284284-64284284 on Build GRCh38
rs779696083 Chr.11: 64284145-64284145 on Build GRCh38
rs754397350 Chr.11: 64284233-64284233 on Build GRCh38
rs758133440 Chr.11: 64284235-64284235 on Build GRCh38
rs561233290 Chr.11: 64284310-64284310 on Build GRCh38
rs142277603 Chr.11: 64284281-64284281 on Build GRCh38

View Full Product Details