Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43592064-43592293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGQ
Entrez Gene ID
126298
Gene Name
immunity related GTPase Q
Gene Aliases
FKSG27, IRGQ1
UniGene
Hs.6217
Gene Chromosome Location
Chr.19: 43584367-43596234 on Build GRCh38
Transcripts
NM_001007561.2 XM_005258515.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
245058 c.1833T>C p.D611D Chr.19: 43592065-43592065 on Build GRCh38
6151604 c.1750G>T p.G584C Chr.19: 43592148-43592148 on Build GRCh38
3535287 c.1794C>T p.H598H Chr.19: 43592104-43592104 on Build GRCh38
4393629 c.1826G>A p.R609Q Chr.19: 43592072-43592072 on Build GRCh38
3535288 c.1737C>T p.F579F Chr.19: 43592161-43592161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772662706 Chr.19: 43592149-43592149 on Build GRCh38
rs753202129 Chr.19: 43592200-43592200 on Build GRCh38
rs868388421 Chr.19: 43592217-43592217 on Build GRCh38
rs753288288 Chr.19: 43592091-43592091 on Build GRCh38
rs184001473 Chr.19: 43592138-43592138 on Build GRCh38
rs757466765 Chr.19: 43592237-43592237 on Build GRCh38
rs769675942 Chr.19: 43592266-43592266 on Build GRCh38
rs756588498 Chr.19: 43592211-43592211 on Build GRCh38
rs756173333 Chr.19: 43592097-43592097 on Build GRCh38
rs754705881 Chr.19: 43592252-43592252 on Build GRCh38
rs762652606 Chr.19: 43592179-43592179 on Build GRCh38
rs748790439 Chr.19: 43592270-43592270 on Build GRCh38
rs768016865 Chr.19: 43592196-43592196 on Build GRCh38
rs796602118 Chr.19: 43592111-43592111 on Build GRCh38
rs759939550 Chr.19: 43592188-43592188 on Build GRCh38
rs746843965 Chr.19: 43592120-43592120 on Build GRCh38
rs199863268 Chr.19: 43592215-43592215 on Build GRCh38
rs746106745 Chr.19: 43592240-43592240 on Build GRCh38
rs748048278 Chr.19: 43592137-43592137 on Build GRCh38
rs774828592 Chr.19: 43592187-43592187 on Build GRCh38
rs145816982 Chr.19: 43592083-43592083 on Build GRCh38
rs567896488 Chr.19: 43592238-43592238 on Build GRCh38
rs768466106 Chr.19: 43592134-43592134 on Build GRCh38
rs776498754 Chr.19: 43592136-43592136 on Build GRCh38
rs193920821 Chr.19: 43592103-43592103 on Build GRCh38
rs748112584 Chr.19: 43592265-43592265 on Build GRCh38
rs757490738 Chr.19: 43592114-43592114 on Build GRCh38
rs780969464 Chr.19: 43592256-43592256 on Build GRCh38
rs772755648 Chr.19: 43592269-43592269 on Build GRCh38
rs749463187 Chr.19: 43592110-43592110 on Build GRCh38
rs765997800 Chr.19: 43592185-43592185 on Build GRCh38
rs913054114 Chr.19: 43592092-43592092 on Build GRCh38
rs900122121 Chr.19: 43592085-43592085 on Build GRCh38
rs912488653 Chr.19: 43592233-43592233 on Build GRCh38
rs753926514 Chr.19: 43592235-43592235 on Build GRCh38
rs779799792 Chr.19: 43592118-43592118 on Build GRCh38
rs769280483 Chr.19: 43592143-43592143 on Build GRCh38
rs1020689328 Chr.19: 43592203-43592203 on Build GRCh38

View Full Product Details