Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53774057-53774284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

Gene Symbol
CHAC2
Entrez Gene ID
494143
Gene Name
ChaC cation transport regulator homolog 2
Gene Aliases
-
UniGene
Hs.585944
Gene Chromosome Location
Chr.2: 53767792-53775150 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5651338 c.263G>A p.R88K Chr.2: 53774233-53774233 on Build GRCh38
1632043 c.181T>C p.W61R Chr.2: 53774151-53774151 on Build GRCh38
6158389 c.217G>A p.E73K Chr.2: 53774187-53774187 on Build GRCh38
1021571 c.240C>T p.D80D Chr.2: 53774210-53774210 on Build GRCh38
239305 c.229G>C p.A77P Chr.2: 53774199-53774199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753253623 Chr.2: 53774184-53774186 on Build GRCh38
rs17851583 Chr.2: 53774214-53774214 on Build GRCh38
rs779500943 Chr.2: 53774182-53774182 on Build GRCh38
rs758890369 Chr.2: 53774168-53774168 on Build GRCh38
rs770450188 Chr.2: 53774138-53774139 on Build GRCh38
rs768113426 Chr.2: 53774164-53774165 on Build GRCh38
rs767331700 Chr.2: 53774110-53774110 on Build GRCh38
rs759809478 Chr.2: 53774236-53774236 on Build GRCh38
rs755017359 Chr.2: 53774148-53774148 on Build GRCh38
rs753872446 Chr.2: 53774143-53774143 on Build GRCh38
rs759800484 Chr.2: 53774143-53774144 on Build GRCh38
rs765351360 Chr.2: 53774242-53774242 on Build GRCh38
rs776809846 Chr.2: 53774226-53774226 on Build GRCh38
rs762673202 Chr.2: 53774122-53774126 on Build GRCh38
rs761705587 Chr.2: 53774108-53774108 on Build GRCh38
rs750189889 Chr.2: 53774111-53774111 on Build GRCh38
rs559346880 Chr.2: 53774097-53774097 on Build GRCh38
rs753231743 Chr.2: 53774155-53774155 on Build GRCh38
rs374101362 Chr.2: 53774191-53774193 on Build GRCh38
rs747351402 Chr.2: 53774172-53774172 on Build GRCh38
rs773010269 Chr.2: 53774213-53774213 on Build GRCh38
rs771917457 Chr.2: 53774208-53774208 on Build GRCh38
rs138719259 Chr.2: 53774240-53774240 on Build GRCh38
rs752720305 Chr.2: 53774252-53774252 on Build GRCh38
rs768676498 Chr.2: 53774103-53774103 on Build GRCh38
rs766200132 Chr.2: 53774224-53774224 on Build GRCh38
rs901229310 Chr.2: 53774091-53774091 on Build GRCh38
rs773792889 Chr.2: 53774196-53774196 on Build GRCh38
rs760827663 Chr.2: 53774113-53774113 on Build GRCh38
rs201851440 Chr.2: 53774093-53774093 on Build GRCh38
rs766609404 Chr.2: 53774125-53774125 on Build GRCh38
rs373573897 Chr.2: 53774191-53774191 on Build GRCh38
rs758886650 Chr.2: 53774255-53774255 on Build GRCh38
rs528086890 Chr.2: 53774099-53774099 on Build GRCh38
rs768260163 Chr.2: 53774193-53774193 on Build GRCh38
rs371632565 Chr.2: 53774186-53774186 on Build GRCh38
rs145056139 Chr.2: 53774170-53774170 on Build GRCh38
rs367818994 Chr.2: 53774217-53774217 on Build GRCh38
rs774801397 Chr.2: 53774140-53774143 on Build GRCh38
rs548342389 Chr.2: 53774107-53774107 on Build GRCh38
rs764535196 Chr.2: 53774256-53774256 on Build GRCh38
rs778136759 Chr.2: 53774169-53774169 on Build GRCh38
rs748112762 Chr.2: 53774204-53774204 on Build GRCh38

View Full Product Details