Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46346854-46347113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPAS1
Entrez Gene ID
2034
Gene Name
endothelial PAS domain protein 1
Gene Aliases
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73
UniGene
Hs.468410
Gene Chromosome Location
Chr.2: 46297402-46386703 on Build GRCh38
Transcripts
NM_001430.4 XM_011532698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681150 c.92C>T p.T31M Chr.2: 46346938-46346938 on Build GRCh38
5643031 c.173G>A p.R58Q Chr.2: 46347019-46347019 on Build GRCh38
5993540 c.69G>A p.A23A Chr.2: 46346915-46346915 on Build GRCh38
442950 c.164C>T p.S55F Chr.2: 46347010-46347010 on Build GRCh38
6997099 c.93G>A p.T31T Chr.2: 46346939-46346939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780355908 Chr.2: 46347045-46347045 on Build GRCh38
rs370020998 Chr.2: 46346939-46346939 on Build GRCh38
rs536627622 Chr.2: 46346878-46346878 on Build GRCh38
rs759108016 Chr.2: 46346945-46346945 on Build GRCh38
rs769385664 Chr.2: 46347091-46347091 on Build GRCh38
rs781102502 Chr.2: 46347034-46347034 on Build GRCh38
rs566608329 Chr.2: 46346978-46346978 on Build GRCh38
rs774946498 Chr.2: 46346936-46346936 on Build GRCh38
rs750515166 Chr.2: 46347038-46347038 on Build GRCh38
rs764991841 Chr.2: 46346954-46346954 on Build GRCh38
rs376889179 Chr.2: 46346881-46346881 on Build GRCh38
rs768863978 Chr.2: 46346913-46346913 on Build GRCh38
rs779189080 Chr.2: 46347068-46347068 on Build GRCh38
rs370989223 Chr.2: 46347080-46347080 on Build GRCh38
rs149032648 Chr.2: 46347053-46347053 on Build GRCh38
rs374587818 Chr.2: 46347039-46347039 on Build GRCh38
rs753774102 Chr.2: 46347073-46347073 on Build GRCh38
rs757224585 Chr.2: 46347027-46347027 on Build GRCh38
rs763745333 Chr.2: 46346991-46346991 on Build GRCh38
rs766269051 Chr.2: 46346979-46346979 on Build GRCh38
rs747537115 Chr.2: 46347094-46347095 on Build GRCh38
rs377637593 Chr.2: 46347077-46347077 on Build GRCh38
rs745372699 Chr.2: 46347086-46347086 on Build GRCh38
rs749668860 Chr.2: 46346912-46346912 on Build GRCh38
rs745916461 Chr.2: 46346882-46346882 on Build GRCh38
rs528826650 Chr.2: 46347008-46347008 on Build GRCh38
rs780192014 Chr.2: 46346909-46346909 on Build GRCh38
rs772679458 Chr.2: 46346942-46346942 on Build GRCh38
rs756121061 Chr.2: 46347040-46347040 on Build GRCh38
rs776351419 Chr.2: 46346943-46346943 on Build GRCh38
rs768904114 Chr.2: 46347058-46347058 on Build GRCh38
rs752221582 Chr.2: 46346973-46346973 on Build GRCh38
rs73926269 Chr.2: 46346887-46346887 on Build GRCh38
rs149898744 Chr.2: 46346992-46346992 on Build GRCh38

View Full Product Details