Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16834835-16835060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
UniGene
Hs.166206
Gene Chromosome Location
Chr.10: 16823966-17130492 on Build GRCh38
Transcripts
NM_001081.3 XM_011519709.2 XM_011519710.2 XM_011519711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702097 c.10351G>A p.D3451N Chr.10: 16835025-16835025 on Build GRCh38
916763 c.10362+2T>C p.? Chr.10: 16835012-16835012 on Build GRCh38
5836628 c.10362+1G>A p.? Chr.10: 16835013-16835013 on Build GRCh38
4675527 c.10337T>C p.V3446A Chr.10: 16835039-16835039 on Build GRCh38
916764 c.10327G>A p.E3443K Chr.10: 16835049-16835049 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750697916 Chr.10: 16834994-16834994 on Build GRCh38
rs703060 Chr.10: 16834923-16834923 on Build GRCh38
rs180968528 Chr.10: 16834924-16834924 on Build GRCh38
rs1034347553 Chr.10: 16835000-16835000 on Build GRCh38
rs943954157 Chr.10: 16834874-16834874 on Build GRCh38
rs186426453 Chr.10: 16834992-16834992 on Build GRCh38
rs772809239 Chr.10: 16835037-16835037 on Build GRCh38
rs547284067 Chr.10: 16834977-16834977 on Build GRCh38
rs749126611 Chr.10: 16835020-16835020 on Build GRCh38
rs747982383 Chr.10: 16835027-16835027 on Build GRCh38
rs771874206 Chr.10: 16835034-16835034 on Build GRCh38
rs145661638 Chr.10: 16835025-16835025 on Build GRCh38
rs778538546 Chr.10: 16835026-16835026 on Build GRCh38
rs564465764 Chr.10: 16834916-16834916 on Build GRCh38
rs1041298849 Chr.10: 16834889-16834889 on Build GRCh38
rs369136480 Chr.10: 16834969-16834969 on Build GRCh38
rs370911251 Chr.10: 16834993-16834993 on Build GRCh38
rs780190067 Chr.10: 16835008-16835008 on Build GRCh38
rs896971404 Chr.10: 16834921-16834921 on Build GRCh38
rs752029852 Chr.10: 16834985-16834985 on Build GRCh38
rs552640113 Chr.10: 16834866-16834866 on Build GRCh38
rs1048432731 Chr.10: 16834946-16834946 on Build GRCh38
rs776574207 Chr.10: 16835007-16835007 on Build GRCh38

View Full Product Details