Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33293953-33294178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGL2
Entrez Gene ID
5863
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Gene Aliases
HKE1.5, KE1.5, RAB2L
UniGene
Hs.509622
Gene Chromosome Location
Chr.6: 33291654-33299388 on Build GRCh38
Transcripts
NM_001243738.1 NM_004761.4 XM_011514781.1 XM_011514785.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765612 c.1147G>A p.G383R Chr.6: 33294027-33294027 on Build GRCh38
4773432 c.1221G>A p.L407L Chr.6: 33293953-33293953 on Build GRCh38
4872213 c.1125T>G p.F375L Chr.6: 33294049-33294049 on Build GRCh38
6765611 c.1386+7G>A p.? Chr.6: 33294027-33294027 on Build GRCh38
3430541 c.1378C>T p.R460W Chr.6: 33294042-33294042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780489836 Chr.6: 33294031-33294031 on Build GRCh38
rs572311254 Chr.6: 33294113-33294113 on Build GRCh38
rs755595818 Chr.6: 33293989-33293989 on Build GRCh38
rs762139070 Chr.6: 33294118-33294119 on Build GRCh38
rs774611599 Chr.6: 33294042-33294042 on Build GRCh38
rs768031311 Chr.6: 33293984-33293984 on Build GRCh38
rs535145420 Chr.6: 33294024-33294024 on Build GRCh38
rs772472633 Chr.6: 33294057-33294057 on Build GRCh38
rs1029733243 Chr.6: 33294120-33294120 on Build GRCh38
rs369080759 Chr.6: 33294019-33294019 on Build GRCh38
rs769980056 Chr.6: 33294030-33294030 on Build GRCh38
rs574507182 Chr.6: 33294075-33294075 on Build GRCh38
rs760249026 Chr.6: 33294070-33294070 on Build GRCh38
rs955468300 Chr.6: 33294138-33294138 on Build GRCh38
rs2239842 Chr.6: 33294094-33294094 on Build GRCh38
rs766081188 Chr.6: 33293992-33293992 on Build GRCh38
rs752485637 Chr.6: 33294012-33294012 on Build GRCh38
rs751761632 Chr.6: 33294119-33294120 on Build GRCh38
rs368176076 Chr.6: 33293985-33293985 on Build GRCh38
rs373999463 Chr.6: 33294103-33294103 on Build GRCh38
rs142317932 Chr.6: 33294028-33294028 on Build GRCh38
rs773758886 Chr.6: 33294060-33294060 on Build GRCh38
rs886421146 Chr.6: 33294151-33294151 on Build GRCh38
rs764901098 Chr.6: 33294095-33294095 on Build GRCh38
rs964552692 Chr.6: 33294108-33294108 on Build GRCh38
rs374811787 Chr.6: 33294006-33294006 on Build GRCh38
rs758153675 Chr.6: 33294112-33294112 on Build GRCh38
rs371599759 Chr.6: 33294003-33294003 on Build GRCh38
rs937736100 Chr.6: 33293986-33293986 on Build GRCh38
rs376652829 Chr.6: 33294071-33294071 on Build GRCh38
rs753488519 Chr.6: 33293998-33293998 on Build GRCh38
rs768952150 Chr.6: 33294041-33294041 on Build GRCh38
rs903767305 Chr.6: 33294092-33294092 on Build GRCh38
rs867176486 Chr.6: 33294027-33294027 on Build GRCh38
rs762250999 Chr.6: 33293973-33293973 on Build GRCh38

View Full Product Details