Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2692606-2692831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUH
Entrez Gene ID
23277
Gene Name
clustered mitochondria homolog
Gene Aliases
CLU1, KIAA0664
UniGene
Hs.22616
Gene Chromosome Location
Chr.17: 2689386-2711633 on Build GRCh38
Transcripts
NM_015229.3 XM_011523768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145826 c.3262G>T p.A1088S Chr.17: 2692630-2692630 on Build GRCh38
1722137 c.3182C>T p.T1061I Chr.17: 2692793-2692793 on Build GRCh38
460400 c.3159G>C p.V1053V Chr.17: 2692816-2692816 on Build GRCh38
4064828 c.3266G>A p.R1089H Chr.17: 2692626-2692626 on Build GRCh38
1255605 c.3261C>T p.R1087R Chr.17: 2692631-2692631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750299603 Chr.17: 2692677-2692677 on Build GRCh38
rs768831760 Chr.17: 2692637-2692637 on Build GRCh38
rs761751463 Chr.17: 2692663-2692663 on Build GRCh38
rs768361755 Chr.17: 2692781-2692781 on Build GRCh38
rs746216542 Chr.17: 2692774-2692774 on Build GRCh38
rs995435238 Chr.17: 2692634-2692634 on Build GRCh38
rs372462099 Chr.17: 2692628-2692628 on Build GRCh38
rs755267780 Chr.17: 2692735-2692735 on Build GRCh38
rs768238950 Chr.17: 2692709-2692709 on Build GRCh38
rs948982858 Chr.17: 2692713-2692713 on Build GRCh38
rs780368280 Chr.17: 2692685-2692685 on Build GRCh38
rs776235025 Chr.17: 2692792-2692792 on Build GRCh38
rs1019154638 Chr.17: 2692722-2692722 on Build GRCh38
rs1029509804 Chr.17: 2692691-2692691 on Build GRCh38
rs898548399 Chr.17: 2692765-2692765 on Build GRCh38
rs34501265 Chr.17: 2692797-2692798 on Build GRCh38
rs928818723 Chr.17: 2692728-2692728 on Build GRCh38
rs765259276 Chr.17: 2692725-2692725 on Build GRCh38
rs763617381 Chr.17: 2692657-2692657 on Build GRCh38
rs1018767226 Chr.17: 2692652-2692652 on Build GRCh38
rs931332739 Chr.17: 2692773-2692773 on Build GRCh38
rs766305548 Chr.17: 2692732-2692732 on Build GRCh38
rs113374324 Chr.17: 2692779-2692779 on Build GRCh38
rs775094871 Chr.17: 2692656-2692656 on Build GRCh38
rs752895732 Chr.17: 2692745-2692745 on Build GRCh38
rs769205501 Chr.17: 2692807-2692807 on Build GRCh38
rs762894650 Chr.17: 2692730-2692730 on Build GRCh38
rs748183821 Chr.17: 2692703-2692703 on Build GRCh38
rs376070458 Chr.17: 2692639-2692639 on Build GRCh38
rs879131326 Chr.17: 2692666-2692666 on Build GRCh38
rs779255901 Chr.17: 2692764-2692764 on Build GRCh38
rs889015605 Chr.17: 2692643-2692643 on Build GRCh38
rs778394080 Chr.17: 2692753-2692753 on Build GRCh38
rs774651555 Chr.17: 2692707-2692707 on Build GRCh38
rs370674871 Chr.17: 2692705-2692705 on Build GRCh38
rs767270498 Chr.17: 2692655-2692655 on Build GRCh38
rs761434867 Chr.17: 2692796-2692796 on Build GRCh38
rs1049689788 Chr.17: 2692784-2692784 on Build GRCh38
rs569372897 Chr.17: 2692758-2692758 on Build GRCh38
rs370497985 Chr.17: 2692649-2692649 on Build GRCh38
rs753306124 Chr.17: 2692660-2692660 on Build GRCh38
rs546501517 Chr.17: 2692708-2692708 on Build GRCh38
rs747122833 Chr.17: 2692630-2692630 on Build GRCh38
rs756120490 Chr.17: 2692751-2692751 on Build GRCh38
rs780676435 Chr.17: 2692771-2692772 on Build GRCh38
rs770395258 Chr.17: 2692704-2692704 on Build GRCh38
rs772733408 Chr.17: 2692808-2692808 on Build GRCh38
rs566688021 Chr.17: 2692711-2692711 on Build GRCh38
rs906449643 Chr.17: 2692648-2692648 on Build GRCh38
rs758702771 Chr.17: 2692627-2692627 on Build GRCh38
rs867006985 Chr.17: 2692646-2692646 on Build GRCh38
rs375661603 Chr.17: 2692724-2692724 on Build GRCh38
rs781423570 Chr.17: 2692701-2692701 on Build GRCh38
rs755208507 Chr.17: 2692700-2692700 on Build GRCh38
rs372755886 Chr.17: 2692651-2692651 on Build GRCh38
rs370490531 Chr.17: 2692752-2692752 on Build GRCh38
rs781554468 Chr.17: 2692741-2692741 on Build GRCh38
rs552683035 Chr.17: 2692729-2692729 on Build GRCh38
rs201306902 Chr.17: 2692759-2692759 on Build GRCh38
rs778261451 Chr.17: 2692690-2692690 on Build GRCh38
rs748692718 Chr.17: 2692642-2692642 on Build GRCh38
rs758096195 Chr.17: 2692684-2692684 on Build GRCh38
rs2302199 Chr.17: 2692670-2692670 on Build GRCh38
rs894788707 Chr.17: 2692625-2692625 on Build GRCh38

View Full Product Details