Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112646395-112646654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
UniGene
Hs.713663
Gene Chromosome Location
Chr.4: 112636964-112657592 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6166282 c.275G>T p.R92I Chr.4: 112646423-112646423 on Build GRCh38
378415 c.276A>T p.R92S Chr.4: 112646424-112646424 on Build GRCh38
4397723 c.279A>T p.A93A Chr.4: 112646427-112646427 on Build GRCh38
5060071 c.344_345insG p.E117fs*6 Chr.4: 112646628-112646629 on Build GRCh38
5865587 c.346G>A p.G116R Chr.4: 112646630-112646630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760828030 Chr.4: 112646597-112646597 on Build GRCh38
rs536015391 Chr.4: 112646492-112646492 on Build GRCh38
rs780942413 Chr.4: 112646499-112646499 on Build GRCh38
rs770919509 Chr.4: 112646561-112646561 on Build GRCh38
rs952099516 Chr.4: 112646626-112646626 on Build GRCh38
rs557181131 Chr.4: 112646497-112646497 on Build GRCh38
rs752090425 Chr.4: 112646442-112646443 on Build GRCh38
rs759607066 Chr.4: 112646431-112646431 on Build GRCh38
rs764897964 Chr.4: 112646421-112646425 on Build GRCh38
rs1043643484 Chr.4: 112646579-112646579 on Build GRCh38
rs745760918 Chr.4: 112646498-112646507 on Build GRCh38
rs12642153 Chr.4: 112646551-112646551 on Build GRCh38
rs555710330 Chr.4: 112646523-112646524 on Build GRCh38
rs999899472 Chr.4: 112646542-112646542 on Build GRCh38
rs200393300 Chr.4: 112646604-112646604 on Build GRCh38
rs574582857 Chr.4: 112646512-112646512 on Build GRCh38
rs761138451 Chr.4: 112646453-112646453 on Build GRCh38
rs749566948 Chr.4: 112646482-112646494 on Build GRCh38
rs779018312 Chr.4: 112646493-112646493 on Build GRCh38
rs1009902869 Chr.4: 112646555-112646555 on Build GRCh38
rs775083711 Chr.4: 112646617-112646618 on Build GRCh38
rs757320103 Chr.4: 112646489-112646494 on Build GRCh38
rs1009237499 Chr.4: 112646563-112646563 on Build GRCh38
rs1056687752 Chr.4: 112646452-112646452 on Build GRCh38
rs1044150303 Chr.4: 112646511-112646511 on Build GRCh38
rs754204087 Chr.4: 112646465-112646465 on Build GRCh38
rs540902339 Chr.4: 112646611-112646611 on Build GRCh38
rs765835996 Chr.4: 112646484-112646484 on Build GRCh38
rs569898184 Chr.4: 112646477-112646478 on Build GRCh38
rs777560247 Chr.4: 112646548-112646548 on Build GRCh38
rs778805827 Chr.4: 112646567-112646567 on Build GRCh38
rs55801142 Chr.4: 112646583-112646583 on Build GRCh38
rs767837318 Chr.4: 112646435-112646435 on Build GRCh38
rs934089365 Chr.4: 112646534-112646534 on Build GRCh38
rs921373520 Chr.4: 112646525-112646525 on Build GRCh38
rs374971073 Chr.4: 112646606-112646606 on Build GRCh38
rs554726477 Chr.4: 112646596-112646596 on Build GRCh38
rs767735112 Chr.4: 112646461-112646463 on Build GRCh38
rs759052037 Chr.4: 112646548-112646550 on Build GRCh38
rs1018843966 Chr.4: 112646578-112646578 on Build GRCh38
rs764394273 Chr.4: 112646455-112646455 on Build GRCh38
rs746004823 Chr.4: 112646574-112646574 on Build GRCh38
rs775548361 Chr.4: 112646445-112646445 on Build GRCh38
rs992256491 Chr.4: 112646467-112646469 on Build GRCh38
rs753135720 Chr.4: 112646464-112646465 on Build GRCh38
rs960482888 Chr.4: 112646615-112646615 on Build GRCh38
rs775598872 Chr.4: 112646591-112646591 on Build GRCh38
rs765464136 Chr.4: 112646474-112646475 on Build GRCh38
rs200626117 Chr.4: 112646602-112646602 on Build GRCh38
rs772013764 Chr.4: 112646585-112646585 on Build GRCh38
rs371322141 Chr.4: 112646494-112646494 on Build GRCh38
rs539197890 Chr.4: 112646475-112646480 on Build GRCh38
rs762417072 Chr.4: 112646483-112646483 on Build GRCh38
rs576858846 Chr.4: 112646619-112646619 on Build GRCh38
rs780467045 Chr.4: 112646545-112646548 on Build GRCh38
rs752327804 Chr.4: 112646503-112646503 on Build GRCh38
rs755583413 Chr.4: 112646446-112646451 on Build GRCh38
rs755746821 Chr.4: 112646546-112646546 on Build GRCh38

View Full Product Details