Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153769161-153769404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNB3
Entrez Gene ID
5365
Gene Name
plexin B3
Gene Aliases
PLEXB3, PLEXR, PLXN6
UniGene
Hs.632833
Gene Chromosome Location
Chr.X: 153764196-153779346 on Build GRCh38
Transcripts
NM_001163257.1 NM_005393.2

SNP Detail

SNP ID Chromosome Location
rs1036115653 Chr.X: 153769194-153769194 on Build GRCh38
rs34012054 Chr.X: 153769233-153769233 on Build GRCh38
rs375860184 Chr.X: 153769298-153769298 on Build GRCh38
rs201176459 Chr.X: 153769270-153769270 on Build GRCh38
rs781832443 Chr.X: 153769239-153769239 on Build GRCh38
rs782809682 Chr.X: 153769199-153769199 on Build GRCh38
rs899042683 Chr.X: 153769381-153769381 on Build GRCh38
rs376423175 Chr.X: 153769190-153769190 on Build GRCh38
rs782210563 Chr.X: 153769320-153769320 on Build GRCh38
rs940411614 Chr.X: 153769358-153769358 on Build GRCh38
rs782664107 Chr.X: 153769341-153769341 on Build GRCh38
rs201696853 Chr.X: 153769232-153769232 on Build GRCh38
rs182040889 Chr.X: 153769365-153769365 on Build GRCh38
rs782297379 Chr.X: 153769346-153769346 on Build GRCh38
rs1029322227 Chr.X: 153769260-153769260 on Build GRCh38
rs369718317 Chr.X: 153769283-153769283 on Build GRCh38
rs112533655 Chr.X: 153769296-153769296 on Build GRCh38
rs376062801 Chr.X: 153769272-153769272 on Build GRCh38
rs916999770 Chr.X: 153769357-153769357 on Build GRCh38
rs189692059 Chr.X: 153769209-153769209 on Build GRCh38
rs113517298 Chr.X: 153769313-153769313 on Build GRCh38
rs1056703036 Chr.X: 153769349-153769349 on Build GRCh38
rs926857709 Chr.X: 153769338-153769338 on Build GRCh38
rs782179254 Chr.X: 153769317-153769317 on Build GRCh38
rs782004288 Chr.X: 153769179-153769179 on Build GRCh38
rs782612543 Chr.X: 153769314-153769314 on Build GRCh38
rs995059798 Chr.X: 153769387-153769387 on Build GRCh38
rs782558882 Chr.X: 153769268-153769268 on Build GRCh38
rs111557364 Chr.X: 153769224-153769224 on Build GRCh38
rs782445788 Chr.X: 153769244-153769244 on Build GRCh38
rs988308708 Chr.X: 153769311-153769311 on Build GRCh38
rs782642582 Chr.X: 153769292-153769292 on Build GRCh38
rs782068722 Chr.X: 153769191-153769191 on Build GRCh38
rs1036061424 Chr.X: 153769375-153769375 on Build GRCh38
rs28447247 Chr.X: 153769226-153769226 on Build GRCh38
rs782453440 Chr.X: 153769336-153769336 on Build GRCh38

View Full Product Details