Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135635833-135636084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HDM1
Entrez Gene ID
23518
Gene Name
R3H domain containing 1
Gene Aliases
R3HDM
UniGene
Hs.412462
Gene Chromosome Location
Chr.2: 135531429-135725270 on Build GRCh38
Transcripts
NM_001282798.1 NM_001282799.1 NM_001282800.1 NM_015361.3 XM_005263626.3 XM_005263627.3 XM_005263628.3 XM_005263629.3 XM_006712389.2 XM_011510870.1 XM_011510871.1 XM_011510872.2 XM_011510873.2 XM_011510874.2 XM_011510875.2 XM_011510876.2 XM_011510877.2 XM_011510878.2 XM_011510879.2 XM_011510881.1 XM_017003718.1 XM_017003719.1 XM_017003720.1 XM_017003721.1 XM_017003722.1 XM_017003723.1 XM_017003724.1 XM_017003725.1 XM_017003726.1 XM_017003727.1 XM_017003728.1 XM_017003729.1 XM_017003730.1 XM_017003731.1 XM_017003732.1 XM_017003733.1 XM_017003734.1 XM_017003735.1 XM_017003736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4085008 c.789T>C p.F263F Chr.2: 135635980-135635980 on Build GRCh38
6761084 c.699-9delT p.? Chr.2: 135635881-135635881 on Build GRCh38
5994289 c.757C>T p.R253C Chr.2: 135635948-135635948 on Build GRCh38
7232290 c.706C>T p.Q236* Chr.2: 135635897-135635897 on Build GRCh38
6761083 c.707A>G p.Q236R Chr.2: 135635898-135635898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773385393 Chr.2: 135635868-135635868 on Build GRCh38
rs769801492 Chr.2: 135636033-135636033 on Build GRCh38
rs745458600 Chr.2: 135635952-135635952 on Build GRCh38
rs886712733 Chr.2: 135635930-135635930 on Build GRCh38
rs747436636 Chr.2: 135635995-135635995 on Build GRCh38
rs760468369 Chr.2: 135636044-135636044 on Build GRCh38
rs935116206 Chr.2: 135636037-135636037 on Build GRCh38
rs756787795 Chr.2: 135635923-135635923 on Build GRCh38
rs780275470 Chr.2: 135635975-135635975 on Build GRCh38
rs529689977 Chr.2: 135636038-135636038 on Build GRCh38
rs767160737 Chr.2: 135635886-135635886 on Build GRCh38
rs769017417 Chr.2: 135636006-135636006 on Build GRCh38
rs1005532972 Chr.2: 135635988-135635988 on Build GRCh38
rs898802420 Chr.2: 135636026-135636026 on Build GRCh38
rs775116763 Chr.2: 135636041-135636041 on Build GRCh38
rs752596478 Chr.2: 135635887-135635887 on Build GRCh38
rs764132258 Chr.2: 135635892-135635892 on Build GRCh38
rs771813361 Chr.2: 135635865-135635870 on Build GRCh38
rs775308434 Chr.2: 135635997-135635998 on Build GRCh38
rs995941868 Chr.2: 135636051-135636051 on Build GRCh38
rs74659439 Chr.2: 135635889-135635889 on Build GRCh38
rs373849492 Chr.2: 135636027-135636027 on Build GRCh38
rs565430350 Chr.2: 135635894-135635894 on Build GRCh38
rs763170211 Chr.2: 135635882-135635882 on Build GRCh38
rs758125129 Chr.2: 135635956-135635956 on Build GRCh38
rs777078092 Chr.2: 135636025-135636025 on Build GRCh38
rs778568401 Chr.2: 135635948-135635948 on Build GRCh38
rs773294872 Chr.2: 135636034-135636034 on Build GRCh38
rs944481319 Chr.2: 135635875-135635881 on Build GRCh38
rs756001488 Chr.2: 135635888-135635888 on Build GRCh38
rs568892505 Chr.2: 135635971-135635971 on Build GRCh38
rs1015944171 Chr.2: 135636024-135636024 on Build GRCh38

View Full Product Details