Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63895393-63895654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARK2
Entrez Gene ID
2011
Gene Name
microtubule affinity regulating kinase 2
Gene Aliases
EMK-1, EMK1, PAR-1, Par-1b, Par1b
UniGene
Hs.567261
Gene Chromosome Location
Chr.11: 63838928-63911020 on Build GRCh38
Transcripts
NM_001039469.2 NM_001163296.1 NM_001163297.1 NM_004954.4 NM_017490.3 XM_011544789.1 XM_011544790.1 XM_011544791.1 XM_011544792.1 XM_011544793.1 XM_011544794.1 XM_011544795.1 XM_011544796.1 XM_011544797.1 XM_011544798.1 XM_017017310.1 XM_017017311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6231607 c.156C>T p.D52D Chr.11: 63895600-63895600 on Build GRCh38
6708333 c.288+1G>C p.? Chr.11: 63895634-63895634 on Build GRCh38
6323456 c.174C>T p.S58S Chr.11: 63895618-63895618 on Build GRCh38
929964 c.189+2T>A p.? Chr.11: 63895635-63895635 on Build GRCh38
7112319 c.171_173delCTC p.S60delS Chr.11: 63895615-63895617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918470523 Chr.11: 63895526-63895526 on Build GRCh38
rs182761359 Chr.11: 63895553-63895553 on Build GRCh38
rs994818481 Chr.11: 63895519-63895519 on Build GRCh38
rs566600964 Chr.11: 63895415-63895415 on Build GRCh38
rs972431812 Chr.11: 63895467-63895467 on Build GRCh38
rs746337941 Chr.11: 63895546-63895546 on Build GRCh38
rs756378548 Chr.11: 63895562-63895562 on Build GRCh38
rs775912141 Chr.11: 63895554-63895554 on Build GRCh38
rs998711424 Chr.11: 63895570-63895570 on Build GRCh38
rs765831400 Chr.11: 63895612-63895612 on Build GRCh38
rs190396547 Chr.11: 63895536-63895536 on Build GRCh38
rs755339467 Chr.11: 63895631-63895632 on Build GRCh38
rs771080526 Chr.11: 63895537-63895537 on Build GRCh38
rs897924848 Chr.11: 63895511-63895511 on Build GRCh38
rs772863897 Chr.11: 63895585-63895585 on Build GRCh38
rs944792825 Chr.11: 63895423-63895423 on Build GRCh38
rs534928733 Chr.11: 63895438-63895438 on Build GRCh38
rs1031814459 Chr.11: 63895588-63895588 on Build GRCh38
rs753363995 Chr.11: 63895618-63895618 on Build GRCh38
rs776856383 Chr.11: 63895545-63895545 on Build GRCh38
rs553836469 Chr.11: 63895527-63895527 on Build GRCh38
rs1036884946 Chr.11: 63895500-63895500 on Build GRCh38
rs758169975 Chr.11: 63895535-63895535 on Build GRCh38
rs556726338 Chr.11: 63895576-63895576 on Build GRCh38
rs763369848 Chr.11: 63895571-63895571 on Build GRCh38
rs1016596030 Chr.11: 63895424-63895424 on Build GRCh38
rs1012874463 Chr.11: 63895543-63895543 on Build GRCh38
rs75565937 Chr.11: 63895485-63895485 on Build GRCh38
rs760324570 Chr.11: 63895609-63895609 on Build GRCh38
rs754529752 Chr.11: 63895627-63895627 on Build GRCh38
rs565878320 Chr.11: 63895413-63895413 on Build GRCh38

View Full Product Details