Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133380373-133380602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAOX
Entrez Gene ID
196743
Gene Name
polyamine oxidase (exo-N4-amino)
Gene Aliases
PAO
UniGene
Hs.501578
Gene Chromosome Location
Chr.10: 133379237-133391696 on Build GRCh38
Transcripts
NM_152911.3 NM_207127.2 NM_207128.2 NR_109763.1 NR_109764.1 NR_109765.1 NR_109766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1219219 c.631T>C p.Y211H Chr.10: 133380448-133380448 on Build GRCh38
5360063 c.586G>A p.G196S Chr.10: 133380403-133380403 on Build GRCh38
5175478 c.644C>T p.P215L Chr.10: 133380461-133380461 on Build GRCh38
6128566 c.578G>C p.C193S Chr.10: 133380395-133380395 on Build GRCh38
117435 c.623T>C p.F208S Chr.10: 133380440-133380440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576772265 Chr.10: 133380558-133380558 on Build GRCh38
rs767581726 Chr.10: 133380476-133380476 on Build GRCh38
rs770431068 Chr.10: 133380453-133380453 on Build GRCh38
rs778557624 Chr.10: 133380452-133380452 on Build GRCh38
rs770377859 Chr.10: 133380533-133380533 on Build GRCh38
rs377453084 Chr.10: 133380510-133380510 on Build GRCh38
rs554058983 Chr.10: 133380540-133380540 on Build GRCh38
rs750666184 Chr.10: 133380489-133380489 on Build GRCh38
rs1041251892 Chr.10: 133380474-133380474 on Build GRCh38
rs775112564 Chr.10: 133380471-133380471 on Build GRCh38
rs1002358563 Chr.10: 133380505-133380505 on Build GRCh38
rs779819334 Chr.10: 133380430-133380430 on Build GRCh38
rs1033940132 Chr.10: 133380555-133380555 on Build GRCh38
rs754022009 Chr.10: 133380492-133380492 on Build GRCh38
rs117600655 Chr.10: 133380441-133380441 on Build GRCh38
rs771619317 Chr.10: 133380431-133380431 on Build GRCh38
rs780644894 Chr.10: 133380402-133380402 on Build GRCh38
rs748587087 Chr.10: 133380415-133380415 on Build GRCh38
rs985375203 Chr.10: 133380418-133380418 on Build GRCh38
rs766590628 Chr.10: 133380491-133380491 on Build GRCh38
rs373985741 Chr.10: 133380506-133380506 on Build GRCh38
rs374398436 Chr.10: 133380403-133380403 on Build GRCh38
rs779540938 Chr.10: 133380508-133380508 on Build GRCh38
rs967272420 Chr.10: 133380568-133380568 on Build GRCh38
rs769816764 Chr.10: 133380462-133380462 on Build GRCh38
rs746463704 Chr.10: 133380449-133380449 on Build GRCh38
rs1025591633 Chr.10: 133380572-133380572 on Build GRCh38
rs759488830 Chr.10: 133380461-133380461 on Build GRCh38
rs760799957 Chr.10: 133380490-133380490 on Build GRCh38
rs371934556 Chr.10: 133380511-133380511 on Build GRCh38
rs762868056 Chr.10: 133380475-133380475 on Build GRCh38
rs916886161 Chr.10: 133380560-133380560 on Build GRCh38
rs368895466 Chr.10: 133380557-133380557 on Build GRCh38
rs952161827 Chr.10: 133380498-133380498 on Build GRCh38
rs779127252 Chr.10: 133380411-133380411 on Build GRCh38
rs112773825 Chr.10: 133380509-133380509 on Build GRCh38
rs56389865 Chr.10: 133380534-133380534 on Build GRCh38
rs776509113 Chr.10: 133380460-133380460 on Build GRCh38

View Full Product Details