Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47853019-47853254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
UniGene
Hs.517949
Gene Chromosome Location
Chr.3: 47850690-48089279 on Build GRCh38
Transcripts
NM_001134364.1 NM_002375.4 XM_005265133.4 XM_005265134.4 XM_005265155.4 XM_005265157.4 XM_005265158.2 XM_006713146.3 XM_006713147.3 XM_006713152.3 XM_011533703.2 XM_011533704.2 XM_011533705.1 XM_011533708.2 XM_011533709.2 XM_011533710.2 XM_011533718.1 XM_011533719.2 XM_017006391.1 XM_017006392.1 XM_017006393.1 XM_017006394.1 XM_017006395.1 XM_017006396.1 XM_017006397.1 XM_017006398.1 XM_017006399.1 XM_017006400.1 XM_017006401.1 XM_017006402.1 XM_017006403.1 XM_017006404.1 XM_017006405.1 XM_017006406.1 XM_017006407.1 XM_017006408.1 XM_017006409.1 XM_017006410.1 XM_017006411.1 XM_017006412.1 XM_017006413.1 XM_017006414.1 XM_017006415.1 XM_017006416.1 XM_017006417.1 XM_017006418.1 XM_017006419.1 XM_017006420.1 XM_017006421.1 XM_017006422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180751 c.3423G>C p.Q1141H Chr.3: 47853191-47853191 on Build GRCh38
7180752 c.2628G>C p.Q876H Chr.3: 47853191-47853191 on Build GRCh38
7095814 c.2580C>T p.L860L Chr.3: 47853239-47853239 on Build GRCh38
1242491 c.3435C>T p.S1145S Chr.3: 47853179-47853179 on Build GRCh38
7095813 c.3375C>T p.L1125L Chr.3: 47853239-47853239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771196825 Chr.3: 47853147-47853147 on Build GRCh38
rs948855587 Chr.3: 47853110-47853110 on Build GRCh38
rs764528324 Chr.3: 47853079-47853079 on Build GRCh38
rs113014719 Chr.3: 47853128-47853128 on Build GRCh38
rs776098747 Chr.3: 47853075-47853075 on Build GRCh38
rs895981242 Chr.3: 47853099-47853099 on Build GRCh38
rs745990474 Chr.3: 47853178-47853178 on Build GRCh38
rs749975072 Chr.3: 47853119-47853119 on Build GRCh38
rs145813357 Chr.3: 47853232-47853232 on Build GRCh38
rs891736238 Chr.3: 47853221-47853221 on Build GRCh38
rs200974649 Chr.3: 47853065-47853065 on Build GRCh38
rs769205952 Chr.3: 47853194-47853194 on Build GRCh38
rs750016496 Chr.3: 47853230-47853230 on Build GRCh38
rs761246042 Chr.3: 47853192-47853192 on Build GRCh38
rs772011572 Chr.3: 47853183-47853183 on Build GRCh38
rs112129588 Chr.3: 47853162-47853162 on Build GRCh38
rs535600402 Chr.3: 47853213-47853213 on Build GRCh38
rs754826125 Chr.3: 47853130-47853130 on Build GRCh38
rs774492719 Chr.3: 47853054-47853054 on Build GRCh38
rs747869687 Chr.3: 47853146-47853146 on Build GRCh38
rs746927973 Chr.3: 47853126-47853126 on Build GRCh38
rs201677916 Chr.3: 47853049-47853049 on Build GRCh38
rs770983785 Chr.3: 47853051-47853051 on Build GRCh38
rs1051418702 Chr.3: 47853083-47853083 on Build GRCh38
rs778951556 Chr.3: 47853165-47853165 on Build GRCh38
rs753598462 Chr.3: 47853081-47853081 on Build GRCh38
rs764975071 Chr.3: 47853229-47853229 on Build GRCh38
rs776185193 Chr.3: 47853189-47853189 on Build GRCh38
rs780081738 Chr.3: 47853122-47853122 on Build GRCh38
rs760206022 Chr.3: 47853217-47853218 on Build GRCh38
rs376401535 Chr.3: 47853121-47853121 on Build GRCh38
rs1044127294 Chr.3: 47853111-47853111 on Build GRCh38
rs759475941 Chr.3: 47853061-47853061 on Build GRCh38
rs762038599 Chr.3: 47853214-47853214 on Build GRCh38
rs756961864 Chr.3: 47853112-47853112 on Build GRCh38

View Full Product Details