Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75218349-75218622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A5
Entrez Gene ID
204962
Gene Name
solute carrier family 44 member 5
Gene Aliases
CTL5
UniGene
Hs.654821
Gene Chromosome Location
Chr.1: 75202131-75724261 on Build GRCh38
Transcripts
NM_001130058.1 NM_001320283.1 NM_001320285.1 NM_001320287.1 NM_152697.4 XM_006710445.3 XM_011540986.2 XM_011540987.2 XM_017000607.1 XM_017000608.1 XM_017000609.1 XM_017000610.1 XM_017000611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6782386 c.1459T>C p.Y487H Chr.1: 75218560-75218560 on Build GRCh38
5678745 c.1488C>T p.I496I Chr.1: 75218531-75218531 on Build GRCh38
2241828 c.1420T>C p.L474L Chr.1: 75218599-75218599 on Build GRCh38
3706030 c.1467C>A p.A489A Chr.1: 75218552-75218552 on Build GRCh38
1265944 c.1424G>A p.G475D Chr.1: 75218595-75218595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756961911 Chr.1: 75218439-75218439 on Build GRCh38
rs745316048 Chr.1: 75218445-75218445 on Build GRCh38
rs267598721 Chr.1: 75218553-75218553 on Build GRCh38
rs201045073 Chr.1: 75218570-75218570 on Build GRCh38
rs531820357 Chr.1: 75218522-75218522 on Build GRCh38
rs750135672 Chr.1: 75218517-75218517 on Build GRCh38
rs757126968 Chr.1: 75218507-75218507 on Build GRCh38
rs748790514 Chr.1: 75218535-75218535 on Build GRCh38
rs138689403 Chr.1: 75218549-75218549 on Build GRCh38
rs779596124 Chr.1: 75218530-75218530 on Build GRCh38
rs779346781 Chr.1: 75218542-75218542 on Build GRCh38
rs764001384 Chr.1: 75218501-75218501 on Build GRCh38
rs1022487842 Chr.1: 75218482-75218482 on Build GRCh38
rs772456359 Chr.1: 75218558-75218558 on Build GRCh38
rs774748209 Chr.1: 75218468-75218468 on Build GRCh38
rs112204236 Chr.1: 75218471-75218471 on Build GRCh38
rs200948281 Chr.1: 75218500-75218500 on Build GRCh38
rs780388201 Chr.1: 75218462-75218462 on Build GRCh38
rs769202811 Chr.1: 75218536-75218536 on Build GRCh38
rs773428036 Chr.1: 75218491-75218491 on Build GRCh38
rs1035423901 Chr.1: 75218527-75218527 on Build GRCh38
rs781045637 Chr.1: 75218441-75218441 on Build GRCh38
rs753271100 Chr.1: 75218490-75218490 on Build GRCh38
rs1026614358 Chr.1: 75218443-75218443 on Build GRCh38
rs148196192 Chr.1: 75218588-75218588 on Build GRCh38
rs779362566 Chr.1: 75218454-75218454 on Build GRCh38
rs748454522 Chr.1: 75218546-75218546 on Build GRCh38
rs769094847 Chr.1: 75218465-75218465 on Build GRCh38
rs143004355 Chr.1: 75218499-75218499 on Build GRCh38
rs370214792 Chr.1: 75218580-75218580 on Build GRCh38
rs72682021 Chr.1: 75218417-75218417 on Build GRCh38
rs769436884 Chr.1: 75218449-75218449 on Build GRCh38
rs751383141 Chr.1: 75218504-75218504 on Build GRCh38
rs780929295 Chr.1: 75218508-75218508 on Build GRCh38
rs773373016 Chr.1: 75218569-75218569 on Build GRCh38
rs759590084 Chr.1: 75218587-75218587 on Build GRCh38
rs144034767 Chr.1: 75218495-75218495 on Build GRCh38
rs755639803 Chr.1: 75218518-75218518 on Build GRCh38

View Full Product Details