Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65671838-65672087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGTB
Entrez Gene ID
54557
Gene Name
small glutamine rich tetratricopeptide repeat containing beta
Gene Aliases
SGT2
UniGene
Hs.482301
Gene Chromosome Location
Chr.5: 65665928-65722821 on Build GRCh38
Transcripts
NM_019072.2 XM_005248548.3 XM_017009597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4558357 c.763G>A p.G255R Chr.5: 65671955-65671955 on Build GRCh38
5403715 c.727G>A p.G243R Chr.5: 65671991-65671991 on Build GRCh38
3855898 c.765A>C p.G255G Chr.5: 65671953-65671953 on Build GRCh38
5455420 c.776T>C p.L259P Chr.5: 65671942-65671942 on Build GRCh38
5004392 c.744C>T p.A248A Chr.5: 65671974-65671974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959972611 Chr.5: 65671923-65671923 on Build GRCh38
rs376391253 Chr.5: 65671908-65671908 on Build GRCh38
rs745622506 Chr.5: 65671956-65671956 on Build GRCh38
rs73763181 Chr.5: 65672022-65672022 on Build GRCh38
rs774006877 Chr.5: 65672021-65672021 on Build GRCh38
rs112879488 Chr.5: 65672050-65672050 on Build GRCh38
rs778969411 Chr.5: 65671890-65671890 on Build GRCh38
rs750129931 Chr.5: 65672040-65672040 on Build GRCh38
rs748311630 Chr.5: 65671901-65671901 on Build GRCh38
rs1034116297 Chr.5: 65671906-65671906 on Build GRCh38
rs568325875 Chr.5: 65671965-65671965 on Build GRCh38
rs998325091 Chr.5: 65671911-65671911 on Build GRCh38
rs767068984 Chr.5: 65672029-65672029 on Build GRCh38
rs769681243 Chr.5: 65671970-65671970 on Build GRCh38
rs763626419 Chr.5: 65672006-65672006 on Build GRCh38
rs375891747 Chr.5: 65671865-65671866 on Build GRCh38
rs200641132 Chr.5: 65671878-65671878 on Build GRCh38
rs985694690 Chr.5: 65672033-65672033 on Build GRCh38
rs370243228 Chr.5: 65671975-65671975 on Build GRCh38
rs979911706 Chr.5: 65671859-65671859 on Build GRCh38
rs775245555 Chr.5: 65671972-65671972 on Build GRCh38
rs945370229 Chr.5: 65671978-65671978 on Build GRCh38
rs1030239998 Chr.5: 65671952-65671952 on Build GRCh38
rs978079701 Chr.5: 65672018-65672018 on Build GRCh38
rs536989518 Chr.5: 65671932-65671932 on Build GRCh38
rs897322446 Chr.5: 65671909-65671909 on Build GRCh38
rs992694001 Chr.5: 65671939-65671939 on Build GRCh38

View Full Product Details