Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159161451-159161708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF145
Entrez Gene ID
153830
Gene Name
ring finger protein 145
Gene Aliases
-
UniGene
Hs.349306
Gene Chromosome Location
Chr.5: 159157409-159210053 on Build GRCh38
Transcripts
NM_001199380.1 NM_001199381.1 NM_001199382.1 NM_001199383.1 NM_144726.2 XM_017009138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395752 c.1376A>T p.Y459F Chr.5: 159161600-159161600 on Build GRCh38
4943308 c.1495G>A p.A499T Chr.5: 159161481-159161481 on Build GRCh38
6389857 c.1378G>T p.V460F Chr.5: 159161598-159161598 on Build GRCh38
3853261 c.1394A>C p.E465A Chr.5: 159161582-159161582 on Build GRCh38
6856401 c.1354-7_1354-4delTTTT p.? Chr.5: 159161626-159161629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532619291 Chr.5: 159161667-159161667 on Build GRCh38
rs773559436 Chr.5: 159161557-159161557 on Build GRCh38
rs138109696 Chr.5: 159161473-159161473 on Build GRCh38
rs763336053 Chr.5: 159161575-159161575 on Build GRCh38
rs753180463 Chr.5: 159161614-159161614 on Build GRCh38
rs759909130 Chr.5: 159161602-159161602 on Build GRCh38
rs766786617 Chr.5: 159161581-159161581 on Build GRCh38
rs753482431 Chr.5: 159161641-159161641 on Build GRCh38
rs763652016 Chr.5: 159161636-159161636 on Build GRCh38
rs780019659 Chr.5: 159161662-159161662 on Build GRCh38
rs530466069 Chr.5: 159161585-159161585 on Build GRCh38
rs533689653 Chr.5: 159161641-159161641 on Build GRCh38
rs76548644 Chr.5: 159161630-159161630 on Build GRCh38
rs930870324 Chr.5: 159161648-159161648 on Build GRCh38
rs552772641 Chr.5: 159161668-159161668 on Build GRCh38
rs374061231 Chr.5: 159161491-159161491 on Build GRCh38
rs750265237 Chr.5: 159161660-159161660 on Build GRCh38
rs748229151 Chr.5: 159161542-159161542 on Build GRCh38
rs200947571 Chr.5: 159161510-159161510 on Build GRCh38
rs757032286 Chr.5: 159161497-159161497 on Build GRCh38
rs370946043 Chr.5: 159161611-159161611 on Build GRCh38
rs773623177 Chr.5: 159161636-159161641 on Build GRCh38
rs780382744 Chr.5: 159161625-159161629 on Build GRCh38
rs565121121 Chr.5: 159161645-159161645 on Build GRCh38
rs987565868 Chr.5: 159161563-159161563 on Build GRCh38
rs59997682 Chr.5: 159161636-159161640 on Build GRCh38
rs758167832 Chr.5: 159161661-159161661 on Build GRCh38
rs772627933 Chr.5: 159161625-159161626 on Build GRCh38
rs769174313 Chr.5: 159161626-159161626 on Build GRCh38
rs770716803 Chr.5: 159161626-159161627 on Build GRCh38
rs374310644 Chr.5: 159161646-159161646 on Build GRCh38
rs747630902 Chr.5: 159161626-159161631 on Build GRCh38
rs367907107 Chr.5: 159161528-159161528 on Build GRCh38
rs201485871 Chr.5: 159161501-159161501 on Build GRCh38
rs371278772 Chr.5: 159161485-159161485 on Build GRCh38
rs758277636 Chr.5: 159161627-159161631 on Build GRCh38
rs778833400 Chr.5: 159161516-159161516 on Build GRCh38
rs1048033967 Chr.5: 159161651-159161651 on Build GRCh38
rs201369350 Chr.5: 159161493-159161493 on Build GRCh38
rs745724719 Chr.5: 159161518-159161518 on Build GRCh38
rs540581579 Chr.5: 159161595-159161595 on Build GRCh38
rs760179941 Chr.5: 159161626-159161629 on Build GRCh38
rs1020060984 Chr.5: 159161490-159161490 on Build GRCh38
rs988702595 Chr.5: 159161489-159161489 on Build GRCh38
rs139682876 Chr.5: 159161626-159161630 on Build GRCh38

View Full Product Details