Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94824459-94824684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
UniGene
Hs.483423
Gene Chromosome Location
Chr.1: 94820342-94895247 on Build GRCh38
Transcripts
NM_001114106.2 NM_001258340.1 NM_001258341.1 NM_001258342.1 NM_001258343.1 NM_001301079.1 NM_152369.4 XM_005270440.2 XM_005270441.2 XM_005270442.1 XM_011540632.2 XM_011540633.2 XM_011540635.2 XM_011540636.2 XM_017000262.1 XM_017000263.1 XM_017000264.1 XM_017000265.1 XM_017000266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3866291 c.249T>C p.P83P Chr.1: 94824606-94824606 on Build GRCh38
3369882 c.42C>T p.L14L Chr.1: 94824543-94824543 on Build GRCh38
3866289 c.246C>T p.A82A Chr.1: 94824603-94824603 on Build GRCh38
1345026 c.12G>A p.S4S Chr.1: 94824513-94824513 on Build GRCh38
2260858 c.273delA p.K93fs*6 Chr.1: 94824630-94824630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763719798 Chr.1: 94824536-94824536 on Build GRCh38
rs550887867 Chr.1: 94824564-94824564 on Build GRCh38
rs770325936 Chr.1: 94824594-94824594 on Build GRCh38
rs145608976 Chr.1: 94824508-94824508 on Build GRCh38
rs747073889 Chr.1: 94824523-94824523 on Build GRCh38
rs148935541 Chr.1: 94824563-94824563 on Build GRCh38
rs1019892007 Chr.1: 94824513-94824513 on Build GRCh38
rs773951394 Chr.1: 94824611-94824611 on Build GRCh38
rs373535073 Chr.1: 94824601-94824601 on Build GRCh38
rs192766621 Chr.1: 94824593-94824593 on Build GRCh38
rs149788729 Chr.1: 94824528-94824528 on Build GRCh38
rs766407430 Chr.1: 94824644-94824644 on Build GRCh38
rs554778369 Chr.1: 94824597-94824597 on Build GRCh38
rs188539443 Chr.1: 94824529-94824529 on Build GRCh38
rs755823364 Chr.1: 94824482-94824482 on Build GRCh38
rs753877700 Chr.1: 94824652-94824652 on Build GRCh38
rs772782515 Chr.1: 94824625-94824625 on Build GRCh38
rs922424519 Chr.1: 94824505-94824505 on Build GRCh38
rs746420514 Chr.1: 94824530-94824530 on Build GRCh38
rs761547638 Chr.1: 94824623-94824623 on Build GRCh38
rs780899800 Chr.1: 94824577-94824579 on Build GRCh38
rs778208592 Chr.1: 94824520-94824520 on Build GRCh38
rs778120746 Chr.1: 94824591-94824591 on Build GRCh38
rs758698244 Chr.1: 94824512-94824512 on Build GRCh38
rs757246523 Chr.1: 94824592-94824592 on Build GRCh38
rs771425075 Chr.1: 94824624-94824624 on Build GRCh38
rs760170626 Chr.1: 94824638-94824638 on Build GRCh38
rs766211094 Chr.1: 94824585-94824585 on Build GRCh38
rs562155133 Chr.1: 94824531-94824531 on Build GRCh38
rs200741955 Chr.1: 94824488-94824488 on Build GRCh38
rs917399982 Chr.1: 94824600-94824600 on Build GRCh38
rs374087822 Chr.1: 94824481-94824481 on Build GRCh38
rs756090190 Chr.1: 94824569-94824569 on Build GRCh38
rs748117099 Chr.1: 94824630-94824630 on Build GRCh38
rs754878626 Chr.1: 94824588-94824588 on Build GRCh38
rs868661782 Chr.1: 94824538-94824538 on Build GRCh38
rs763371251 Chr.1: 94824533-94824533 on Build GRCh38
rs369177498 Chr.1: 94824567-94824567 on Build GRCh38
rs774037852 Chr.1: 94824557-94824557 on Build GRCh38
rs759376584 Chr.1: 94824654-94824654 on Build GRCh38

View Full Product Details