Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108623959-108624184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELPLG
Entrez Gene ID
6404
Gene Name
selectin P ligand
Gene Aliases
CD162, CLA, PSGL-1, PSGL1
UniGene
Hs.591014
Gene Chromosome Location
Chr.12: 108621895-108633894 on Build GRCh38
Transcripts
NM_001206609.1 NM_003006.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1358536 c.266G>A p.R89H Chr.12: 108624042-108624042 on Build GRCh38
6490086 c.248T>C p.V83A Chr.12: 108624060-108624060 on Build GRCh38
6045817 c.321C>T p.A107A Chr.12: 108623987-108623987 on Build GRCh38
4146524 c.186G>A p.M62I Chr.12: 108624122-108624122 on Build GRCh38
4545416 c.329G>A p.G110E Chr.12: 108623979-108623979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149329031 Chr.12: 108623982-108623982 on Build GRCh38
rs2228315 Chr.12: 108624122-108624122 on Build GRCh38
rs780685795 Chr.12: 108624063-108624063 on Build GRCh38
rs764400289 Chr.12: 108624137-108624137 on Build GRCh38
rs753056495 Chr.12: 108624061-108624061 on Build GRCh38
rs368817681 Chr.12: 108624138-108624138 on Build GRCh38
rs367989066 Chr.12: 108623996-108623996 on Build GRCh38
rs770136272 Chr.12: 108624028-108624028 on Build GRCh38
rs778252622 Chr.12: 108624069-108624069 on Build GRCh38
rs778001582 Chr.12: 108624086-108624086 on Build GRCh38
rs753458286 Chr.12: 108624065-108624065 on Build GRCh38
rs147444406 Chr.12: 108623997-108623997 on Build GRCh38
rs181419331 Chr.12: 108624110-108624110 on Build GRCh38
rs770936396 Chr.12: 108624036-108624036 on Build GRCh38
rs758392715 Chr.12: 108624075-108624075 on Build GRCh38
rs755750730 Chr.12: 108623981-108623981 on Build GRCh38
rs61934088 Chr.12: 108624132-108624132 on Build GRCh38
rs201172154 Chr.12: 108624042-108624042 on Build GRCh38
rs764993825 Chr.12: 108624163-108624163 on Build GRCh38
rs771235385 Chr.12: 108624090-108624090 on Build GRCh38
rs760998976 Chr.12: 108624157-108624157 on Build GRCh38
rs761347379 Chr.12: 108624153-108624153 on Build GRCh38
rs1023054593 Chr.12: 108624149-108624149 on Build GRCh38
rs749995403 Chr.12: 108624071-108624071 on Build GRCh38
rs746015434 Chr.12: 108624100-108624100 on Build GRCh38
rs1018628959 Chr.12: 108624043-108624043 on Build GRCh38
rs773310588 Chr.12: 108624034-108624034 on Build GRCh38
rs774283363 Chr.12: 108624095-108624095 on Build GRCh38
rs772504919 Chr.12: 108624102-108624102 on Build GRCh38
rs768071324 Chr.12: 108624053-108624053 on Build GRCh38
rs753326035 Chr.12: 108623983-108623983 on Build GRCh38
rs780203043 Chr.12: 108624081-108624081 on Build GRCh38
rs746855045 Chr.12: 108624007-108624007 on Build GRCh38
rs768584428 Chr.12: 108624085-108624085 on Build GRCh38
rs746734450 Chr.12: 108624084-108624084 on Build GRCh38
rs763031178 Chr.12: 108624035-108624035 on Build GRCh38
rs563890933 Chr.12: 108624089-108624089 on Build GRCh38
rs530681343 Chr.12: 108623988-108623988 on Build GRCh38
rs760015806 Chr.12: 108624052-108624052 on Build GRCh38
rs371499696 Chr.12: 108623992-108623992 on Build GRCh38
rs764918224 Chr.12: 108624163-108624164 on Build GRCh38
rs1027893227 Chr.12: 108624101-108624101 on Build GRCh38
rs756752918 Chr.12: 108624066-108624066 on Build GRCh38
rs184663566 Chr.12: 108623984-108623984 on Build GRCh38
rs745821124 Chr.12: 108623995-108623995 on Build GRCh38

View Full Product Details