Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41028495-41028766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCMH1
Entrez Gene ID
22955
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases
Scml3
UniGene
Hs.571874
Gene Chromosome Location
Chr.1: 41027199-41242155 on Build GRCh38
Transcripts
NM_001031694.2 NM_001172218.1 NM_001172219.1 NM_001172220.1 NM_001172221.1 NM_001172222.2 NM_012236.3 NR_135092.1 XM_006710462.2 XM_006710464.1 XM_011541032.2 XM_011541033.2 XM_011541034.1 XM_011541035.2 XM_011541036.2 XM_011541039.1 XM_011541043.2 XM_011541044.1 XM_011541045.2 XM_017000698.1 XM_017000699.1 XM_017000700.1 XM_017000701.1 XM_017000702.1 XM_017000703.1 XM_017000704.1 XM_017000705.1 XM_017000706.1 XM_017000707.1 XM_017000708.1 XM_017000709.1 XM_017000710.1 XM_017000711.1 XM_017000712.1 XM_017000713.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001031694.2 NM_001172218.1 NM_001172219.1 NM_001172220.1 NM_001172221.1 NM_001172222.2 NM_012236.3 NR_135092.1 XM_006710462.2 XM_006710464.1 XM_011541032.2 XM_011541033.2 XM_011541034.1 XM_011541035.2 XM_011541036.2 XM_011541039.1 XM_011541043.2 XM_011541044.1 XM_011541045.2 XM_017000698.1 XM_017000699.1 XM_017000700.1 XM_017000701.1 XM_017000702.1 XM_017000703.1 XM_017000704.1 XM_017000705.1 XM_017000706.1 XM_017000707.1 XM_017000708.1 XM_017000709.1 XM_017000710.1 XM_017000711.1 XM_017000712.1 XM_017000713.1 NR_037868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909536 c.1802C>T p.A601V Chr.1: 41028603-41028603 on Build GRCh38
3489637 c.1636C>T p.L546L Chr.1: 41028598-41028598 on Build GRCh38
7103580 c.1546C>T p.L516L Chr.1: 41028688-41028688 on Build GRCh38
6772720 c.1849C>T p.R617C Chr.1: 41028592-41028592 on Build GRCh38
4436589 c.1715G>A p.R572Q Chr.1: 41028690-41028690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773982926 Chr.1: 41028582-41028582 on Build GRCh38
rs773637274 Chr.1: 41028549-41028549 on Build GRCh38
rs200377184 Chr.1: 41028547-41028547 on Build GRCh38
rs188686145 Chr.1: 41028540-41028540 on Build GRCh38
rs991254709 Chr.1: 41028519-41028519 on Build GRCh38
rs367631516 Chr.1: 41028604-41028604 on Build GRCh38
rs369637138 Chr.1: 41028720-41028720 on Build GRCh38
rs752695039 Chr.1: 41028689-41028689 on Build GRCh38
rs376233858 Chr.1: 41028678-41028678 on Build GRCh38
rs754973906 Chr.1: 41028558-41028558 on Build GRCh38
rs267598597 Chr.1: 41028679-41028679 on Build GRCh38
rs747407979 Chr.1: 41028702-41028702 on Build GRCh38
rs748715258 Chr.1: 41028717-41028717 on Build GRCh38
rs763639795 Chr.1: 41028695-41028695 on Build GRCh38
rs546399615 Chr.1: 41028670-41028670 on Build GRCh38
rs192535871 Chr.1: 41028703-41028703 on Build GRCh38
rs749331053 Chr.1: 41028721-41028721 on Build GRCh38
rs767493187 Chr.1: 41028680-41028680 on Build GRCh38
rs776939554 Chr.1: 41028633-41028633 on Build GRCh38
rs759332224 Chr.1: 41028704-41028704 on Build GRCh38
rs11578697 Chr.1: 41028550-41028550 on Build GRCh38
rs778375263 Chr.1: 41028701-41028701 on Build GRCh38
rs758238157 Chr.1: 41028674-41028674 on Build GRCh38
rs376364655 Chr.1: 41028690-41028690 on Build GRCh38
rs752214664 Chr.1: 41028573-41028573 on Build GRCh38
rs763307338 Chr.1: 41028659-41028659 on Build GRCh38
rs990926700 Chr.1: 41028718-41028718 on Build GRCh38
rs751280564 Chr.1: 41028556-41028556 on Build GRCh38
rs764308790 Chr.1: 41028549-41028550 on Build GRCh38
rs766246549 Chr.1: 41028553-41028553 on Build GRCh38
rs753310087 Chr.1: 41028700-41028700 on Build GRCh38
rs769223096 Chr.1: 41028605-41028605 on Build GRCh38
rs770394006 Chr.1: 41028719-41028719 on Build GRCh38
rs916575244 Chr.1: 41028528-41028528 on Build GRCh38
rs541970157 Chr.1: 41028531-41028531 on Build GRCh38
rs770058621 Chr.1: 41028650-41028650 on Build GRCh38
rs533089870 Chr.1: 41028655-41028655 on Build GRCh38
rs146565042 Chr.1: 41028723-41028723 on Build GRCh38
rs143623408 Chr.1: 41028591-41028591 on Build GRCh38
rs561840550 Chr.1: 41028538-41028538 on Build GRCh38
rs545780309 Chr.1: 41028722-41028722 on Build GRCh38
rs761928863 Chr.1: 41028546-41028546 on Build GRCh38
rs371463378 Chr.1: 41028618-41028618 on Build GRCh38
rs957935145 Chr.1: 41028682-41028682 on Build GRCh38
rs374927869 Chr.1: 41028671-41028671 on Build GRCh38
rs572966698 Chr.1: 41028516-41028516 on Build GRCh38
rs748662296 Chr.1: 41028634-41028634 on Build GRCh38
rs759797640 Chr.1: 41028744-41028744 on Build GRCh38
rs780503002 Chr.1: 41028593-41028593 on Build GRCh38
rs963484064 Chr.1: 41028517-41028517 on Build GRCh38
rs139648979 Chr.1: 41028592-41028592 on Build GRCh38
rs1052942463 Chr.1: 41028583-41028583 on Build GRCh38
rs200250307 Chr.1: 41028584-41028584 on Build GRCh38
rs564056155 Chr.1: 41028570-41028570 on Build GRCh38

View Full Product Details