Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226642008-226642269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPKB
Entrez Gene ID
3707
Gene Name
inositol-trisphosphate 3-kinase B
Gene Aliases
IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37
UniGene
Hs.528087
Gene Chromosome Location
Chr.1: 226631690-226739327 on Build GRCh38
Transcripts
NM_002221.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550533 c.2339C>T p.A780V Chr.1: 226642033-226642033 on Build GRCh38
679348 c.2322C>G p.I774M Chr.1: 226642050-226642050 on Build GRCh38
3729173 c.2293C>A p.L765M Chr.1: 226642079-226642079 on Build GRCh38
679349 c.900C>G p.I300M Chr.1: 226642050-226642050 on Build GRCh38
4941353 c.843G>A p.E281E Chr.1: 226642107-226642107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763351019 Chr.1: 226642032-226642032 on Build GRCh38
rs1027338395 Chr.1: 226642215-226642215 on Build GRCh38
rs574594922 Chr.1: 226642210-226642210 on Build GRCh38
rs1057255748 Chr.1: 226642029-226642029 on Build GRCh38
rs140588026 Chr.1: 226642103-226642103 on Build GRCh38
rs961791673 Chr.1: 226642161-226642161 on Build GRCh38
rs369471382 Chr.1: 226642155-226642155 on Build GRCh38
rs374484908 Chr.1: 226642091-226642091 on Build GRCh38
rs763440563 Chr.1: 226642134-226642134 on Build GRCh38
rs764485101 Chr.1: 226642138-226642138 on Build GRCh38
rs778624225 Chr.1: 226642177-226642177 on Build GRCh38
rs776013826 Chr.1: 226642130-226642130 on Build GRCh38
rs953470981 Chr.1: 226642194-226642194 on Build GRCh38
rs766251623 Chr.1: 226642162-226642162 on Build GRCh38
rs200871560 Chr.1: 226642094-226642094 on Build GRCh38
rs534711357 Chr.1: 226642129-226642129 on Build GRCh38
rs200031424 Chr.1: 226642093-226642093 on Build GRCh38
rs775754706 Chr.1: 226642030-226642030 on Build GRCh38
rs936616632 Chr.1: 226642230-226642230 on Build GRCh38
rs750503076 Chr.1: 226642050-226642050 on Build GRCh38
rs796260732 Chr.1: 226642216-226642216 on Build GRCh38
rs377329491 Chr.1: 226642163-226642163 on Build GRCh38
rs925314859 Chr.1: 226642218-226642218 on Build GRCh38
rs746493731 Chr.1: 226642118-226642118 on Build GRCh38
rs1002299139 Chr.1: 226642132-226642132 on Build GRCh38
rs377575333 Chr.1: 226642217-226642217 on Build GRCh38
rs923380426 Chr.1: 226642193-226642193 on Build GRCh38
rs751788436 Chr.1: 226642038-226642038 on Build GRCh38
rs747734824 Chr.1: 226642101-226642101 on Build GRCh38
rs372972707 Chr.1: 226642156-226642156 on Build GRCh38
rs771586213 Chr.1: 226642102-226642102 on Build GRCh38
rs764431826 Chr.1: 226642037-226642037 on Build GRCh38
rs780011409 Chr.1: 226642086-226642086 on Build GRCh38
rs774726662 Chr.1: 226642143-226642143 on Build GRCh38
rs767646058 Chr.1: 226642049-226642049 on Build GRCh38
rs757416349 Chr.1: 226642039-226642039 on Build GRCh38
rs761000219 Chr.1: 226642171-226642180 on Build GRCh38
rs373345228 Chr.1: 226642148-226642148 on Build GRCh38
rs754775076 Chr.1: 226642166-226642166 on Build GRCh38
rs749049375 Chr.1: 226642133-226642133 on Build GRCh38
rs41268735 Chr.1: 226642144-226642144 on Build GRCh38

View Full Product Details