Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69455301-69455574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPLP1
Entrez Gene ID
6176
Gene Name
ribosomal protein lateral stalk subunit P1
Gene Aliases
LP1, P1, RPP1
UniGene
Hs.356502
Gene Chromosome Location
Chr.15: 69452820-69455545 on Build GRCh38
Transcripts
NM_001003.2 NM_213725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582928 c.292A>G p.K98E Chr.15: 69455454-69455454 on Build GRCh38
4056540 c.299A>G p.E100G Chr.15: 69455461-69455461 on Build GRCh38
3711735 c.273G>A p.E91E Chr.15: 69455435-69455435 on Build GRCh38
1374295 c.335delT p.F113fs*>2 Chr.15: 69455497-69455497 on Build GRCh38
1323869 c.309G>C p.E103D Chr.15: 69455471-69455471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1463911 Chr.15: 69455336-69455336 on Build GRCh38
rs1004883858 Chr.15: 69455482-69455482 on Build GRCh38
rs754137512 Chr.15: 69455415-69455418 on Build GRCh38
rs781682538 Chr.15: 69455439-69455439 on Build GRCh38
rs780740996 Chr.15: 69455412-69455412 on Build GRCh38
rs116311376 Chr.15: 69455337-69455337 on Build GRCh38
rs753479085 Chr.15: 69455528-69455528 on Build GRCh38
rs762913884 Chr.15: 69455329-69455330 on Build GRCh38
rs930118598 Chr.15: 69455363-69455363 on Build GRCh38
rs774274098 Chr.15: 69455488-69455488 on Build GRCh38
rs576544725 Chr.15: 69455414-69455415 on Build GRCh38
rs202225518 Chr.15: 69455518-69455518 on Build GRCh38
rs200720703 Chr.15: 69455545-69455545 on Build GRCh38
rs761416059 Chr.15: 69455540-69455540 on Build GRCh38
rs763723146 Chr.15: 69455387-69455387 on Build GRCh38
rs760839391 Chr.15: 69455415-69455415 on Build GRCh38
rs201726748 Chr.15: 69455334-69455334 on Build GRCh38
rs11554438 Chr.15: 69455478-69455478 on Build GRCh38
rs748470409 Chr.15: 69455444-69455444 on Build GRCh38
rs891749437 Chr.15: 69455532-69455532 on Build GRCh38
rs755410929 Chr.15: 69455424-69455424 on Build GRCh38
rs906017074 Chr.15: 69455405-69455405 on Build GRCh38
rs796295422 Chr.15: 69455334-69455336 on Build GRCh38
rs760411129 Chr.15: 69455525-69455525 on Build GRCh38
rs762069154 Chr.15: 69455430-69455432 on Build GRCh38
rs544246404 Chr.15: 69455411-69455411 on Build GRCh38
rs1054280719 Chr.15: 69455493-69455493 on Build GRCh38
rs765659388 Chr.15: 69455510-69455511 on Build GRCh38
rs992380171 Chr.15: 69455368-69455368 on Build GRCh38
rs764191639 Chr.15: 69455415-69455417 on Build GRCh38
rs548004580 Chr.15: 69455527-69455527 on Build GRCh38
rs749351343 Chr.15: 69455468-69455468 on Build GRCh38
rs370055255 Chr.15: 69455483-69455483 on Build GRCh38
rs201427547 Chr.15: 69455512-69455512 on Build GRCh38
rs201272949 Chr.15: 69455452-69455452 on Build GRCh38
rs771001182 Chr.15: 69455336-69455336 on Build GRCh38
rs752047646 Chr.15: 69455416-69455416 on Build GRCh38
rs777960502 Chr.15: 69455466-69455466 on Build GRCh38
rs562950236 Chr.15: 69455465-69455465 on Build GRCh38
rs753253861 Chr.15: 69455406-69455406 on Build GRCh38

View Full Product Details