Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 60544594-60544843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIM2
Entrez Gene ID
5558
Gene Name
primase (DNA) subunit 2
Gene Aliases
PRIM2A, p58
UniGene
Hs.654580
Gene Chromosome Location
Chr.6: 57314805-57646852 on Build GRCh38
Transcripts
NM_000947.4 XM_011514731.2 XM_017011004.1 XM_017011007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3159939 c.1390G>A p.E464K Chr.6: 57646018-57646018 on Build GRCh38
5795271 c.1347T>G p.C449W Chr.6: 57645975-57645975 on Build GRCh38
322814 c.1372G>A p.G458S Chr.6: 57646000-57646000 on Build GRCh38
3159932 c.1341C>A p.F447L Chr.6: 57645969-57645969 on Build GRCh38
5759940 c.1327C>T p.H443Y Chr.6: 57645955-57645955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545164255 Chr.6: 60544640-60544640 on Build GRCh38
rs759580128 Chr.6: 60544819-60544819 on Build GRCh38
rs771346275 Chr.6: 60544817-60544817 on Build GRCh38
rs763199623 Chr.6: 60544780-60544780 on Build GRCh38
rs778583005 Chr.6: 60544754-60544754 on Build GRCh38
rs753221894 Chr.6: 60544794-60544794 on Build GRCh38
rs750026434 Chr.6: 60544755-60544755 on Build GRCh38
rs775430961 Chr.6: 60544735-60544735 on Build GRCh38
rs770479247 Chr.6: 60544691-60544691 on Build GRCh38
rs745364992 Chr.6: 60544696-60544696 on Build GRCh38
rs747446127 Chr.6: 60544679-60544679 on Build GRCh38
rs745420112 Chr.6: 60544731-60544731 on Build GRCh38
rs531704936 Chr.6: 60544737-60544737 on Build GRCh38
rs761878973 Chr.6: 60544709-60544709 on Build GRCh38
rs756467372 Chr.6: 60544752-60544752 on Build GRCh38
rs539010550 Chr.6: 60544775-60544775 on Build GRCh38
rs768872342 Chr.6: 60544680-60544680 on Build GRCh38
rs747935707 Chr.6: 60544770-60544770 on Build GRCh38
rs766524977 Chr.6: 60544746-60544746 on Build GRCh38
rs4294008 Chr.6: 60544818-60544818 on Build GRCh38
rs5001076 Chr.6: 60544782-60544782 on Build GRCh38
rs764404860 Chr.6: 60544797-60544797 on Build GRCh38
rs748071924 Chr.6: 60544808-60544808 on Build GRCh38
rs554333791 Chr.6: 60544783-60544783 on Build GRCh38
rs756913357 Chr.6: 60544727-60544727 on Build GRCh38
rs777651782 Chr.6: 60544811-60544811 on Build GRCh38
rs759653088 Chr.6: 60544788-60544788 on Build GRCh38
rs772060272 Chr.6: 60544700-60544700 on Build GRCh38
rs768534684 Chr.6: 60544765-60544765 on Build GRCh38
rs767035154 Chr.6: 60544715-60544715 on Build GRCh38
rs780694081 Chr.6: 60544677-60544677 on Build GRCh38
rs767946117 Chr.6: 60544792-60544792 on Build GRCh38
rs755399322 Chr.6: 60544721-60544721 on Build GRCh38
rs747901501 Chr.6: 60544687-60544687 on Build GRCh38
rs763176836 Chr.6: 60544745-60544745 on Build GRCh38
rs781042111 Chr.6: 60544806-60544806 on Build GRCh38
rs775371659 Chr.6: 60544703-60544703 on Build GRCh38
rs751098291 Chr.6: 60544804-60544804 on Build GRCh38
rs771610934 Chr.6: 60544740-60544740 on Build GRCh38
rs760508820 Chr.6: 60544704-60544704 on Build GRCh38
rs757991075 Chr.6: 60544803-60544803 on Build GRCh38
rs768054226 Chr.6: 60544749-60544749 on Build GRCh38
rs527493097 Chr.6: 60544658-60544658 on Build GRCh38
rs772896163 Chr.6: 60544800-60544801 on Build GRCh38
rs750084029 Chr.6: 60544798-60544798 on Build GRCh38
rs758919117 Chr.6: 60544676-60544676 on Build GRCh38
rs771545390 Chr.6: 60544733-60544733 on Build GRCh38
rs200464149 Chr.6: 60544739-60544739 on Build GRCh38
rs567784436 Chr.6: 60544713-60544713 on Build GRCh38
rs754480915 Chr.6: 60544805-60544805 on Build GRCh38
rs773598160 Chr.6: 60544777-60544777 on Build GRCh38
rs765373596 Chr.6: 60544710-60544710 on Build GRCh38
rs549453753 Chr.6: 60544690-60544690 on Build GRCh38
rs755381677 Chr.6: 60544748-60544748 on Build GRCh38
rs748983475 Chr.6: 60544725-60544725 on Build GRCh38
rs776235564 Chr.6: 60544738-60544738 on Build GRCh38
rs749080265 Chr.6: 60544812-60544812 on Build GRCh38
rs753065889 Chr.6: 60544750-60544750 on Build GRCh38
rs757937644 Chr.6: 60544759-60544759 on Build GRCh38
rs781343743 Chr.6: 60544723-60544723 on Build GRCh38
rs765623048 Chr.6: 60544741-60544741 on Build GRCh38
rs4307164 Chr.6: 60544729-60544729 on Build GRCh38
rs752114969 Chr.6: 60544747-60544747 on Build GRCh38
rs769714703 Chr.6: 60544810-60544810 on Build GRCh38
rs750589632 Chr.6: 60544711-60544711 on Build GRCh38
rs4294007 Chr.6: 60544763-60544763 on Build GRCh38
rs781186146 Chr.6: 60544766-60544766 on Build GRCh38
rs4294006 Chr.6: 60544692-60544692 on Build GRCh38
rs776947001 Chr.6: 60544708-60544708 on Build GRCh38
rs763749474 Chr.6: 60544705-60544705 on Build GRCh38
rs773538877 Chr.6: 60544744-60544744 on Build GRCh38
rs560131306 Chr.6: 60544649-60544649 on Build GRCh38
rs779476179 Chr.6: 60544761-60544761 on Build GRCh38
rs752061895 Chr.6: 60544718-60544718 on Build GRCh38
rs1052780580 Chr.6: 60544815-60544815 on Build GRCh38

View Full Product Details