Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113235207-113235432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX8
Entrez Gene ID
7849
Gene Name
paired box 8
Gene Aliases
-
UniGene
Hs.469728
Gene Chromosome Location
Chr.2: 113215997-113278950 on Build GRCh38
Transcripts
NM_003466.3 NM_013952.3 NM_013953.3 NM_013992.3 XM_011511790.1 XM_011511791.1 XM_011511792.1 XM_011511793.1 XM_011511794.1 XM_017004878.1 XM_017004879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378957 c.1058G>A p.G353E Chr.2: 113235423-113235423 on Build GRCh38
1398915 c.1051G>A p.V351M Chr.2: 113235430-113235430 on Build GRCh38
4582589 c.1049C>T p.S350F Chr.2: 113235432-113235432 on Build GRCh38
4848337 c.1085C>G p.S362* Chr.2: 113235396-113235396 on Build GRCh38
5760574 c.1087+5G>A p.? Chr.2: 113235389-113235389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111481416 Chr.2: 113235390-113235390 on Build GRCh38
rs1022915989 Chr.2: 113235302-113235302 on Build GRCh38
rs754947490 Chr.2: 113235361-113235361 on Build GRCh38
rs778012744 Chr.2: 113235389-113235389 on Build GRCh38
rs1041647755 Chr.2: 113235350-113235350 on Build GRCh38
rs1039165940 Chr.2: 113235342-113235342 on Build GRCh38
rs989337337 Chr.2: 113235319-113235319 on Build GRCh38
rs908098533 Chr.2: 113235226-113235226 on Build GRCh38
rs772655672 Chr.2: 113235415-113235415 on Build GRCh38
rs774778854 Chr.2: 113235404-113235404 on Build GRCh38
rs931085686 Chr.2: 113235288-113235288 on Build GRCh38
rs748252451 Chr.2: 113235387-113235387 on Build GRCh38
rs944899776 Chr.2: 113235326-113235326 on Build GRCh38
rs945928849 Chr.2: 113235346-113235346 on Build GRCh38
rs866789008 Chr.2: 113235290-113235290 on Build GRCh38
rs959899697 Chr.2: 113235284-113235284 on Build GRCh38
rs978802039 Chr.2: 113235313-113235313 on Build GRCh38
rs866301416 Chr.2: 113235227-113235227 on Build GRCh38
rs911976061 Chr.2: 113235322-113235322 on Build GRCh38
rs956159436 Chr.2: 113235318-113235318 on Build GRCh38
rs751519435 Chr.2: 113235354-113235354 on Build GRCh38
rs1015335017 Chr.2: 113235238-113235238 on Build GRCh38
rs568106732 Chr.2: 113235413-113235413 on Build GRCh38
rs559830742 Chr.2: 113235314-113235314 on Build GRCh38
rs962241851 Chr.2: 113235231-113235231 on Build GRCh38
rs759775701 Chr.2: 113235316-113235316 on Build GRCh38
rs925909398 Chr.2: 113235315-113235315 on Build GRCh38
rs969931372 Chr.2: 113235311-113235311 on Build GRCh38
rs370787030 Chr.2: 113235353-113235353 on Build GRCh38
rs397778746 Chr.2: 113235226-113235227 on Build GRCh38
rs756292699 Chr.2: 113235388-113235388 on Build GRCh38
rs911792530 Chr.2: 113235339-113235339 on Build GRCh38
rs528054716 Chr.2: 113235376-113235376 on Build GRCh38

View Full Product Details