Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37675678-37675943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS1
Entrez Gene ID
3956
Gene Name
galectin 1
Gene Aliases
GAL1, GBP
UniGene
Hs.445351
Gene Chromosome Location
Chr.22: 37675606-37679802 on Build GRCh38
Transcripts
NM_002305.3

SNP Detail

SNP ID Chromosome Location
rs201722376 Chr.22: 37675732-37675732 on Build GRCh38
rs990167873 Chr.22: 37675770-37675771 on Build GRCh38
rs376991752 Chr.22: 37675745-37675745 on Build GRCh38
rs1056672527 Chr.22: 37675845-37675845 on Build GRCh38
rs199824702 Chr.22: 37675731-37675731 on Build GRCh38
rs955559428 Chr.22: 37675751-37675751 on Build GRCh38
rs756085054 Chr.22: 37675885-37675885 on Build GRCh38
rs1016610691 Chr.22: 37675912-37675912 on Build GRCh38
rs948692950 Chr.22: 37675802-37675802 on Build GRCh38
rs376111007 Chr.22: 37675704-37675704 on Build GRCh38
rs962440750 Chr.22: 37675736-37675736 on Build GRCh38
rs907268136 Chr.22: 37675818-37675818 on Build GRCh38
rs531096914 Chr.22: 37675787-37675787 on Build GRCh38
rs775584971 Chr.22: 37675731-37675732 on Build GRCh38
rs914523271 Chr.22: 37675789-37675789 on Build GRCh38
rs939197197 Chr.22: 37675824-37675824 on Build GRCh38
rs370095144 Chr.22: 37675755-37675755 on Build GRCh38
rs536010314 Chr.22: 37675724-37675725 on Build GRCh38
rs924899710 Chr.22: 37675760-37675760 on Build GRCh38
rs957930423 Chr.22: 37675919-37675919 on Build GRCh38
rs377124172 Chr.22: 37675733-37675733 on Build GRCh38
rs200510997 Chr.22: 37675756-37675756 on Build GRCh38
rs905061089 Chr.22: 37675727-37675727 on Build GRCh38
rs752640310 Chr.22: 37675849-37675849 on Build GRCh38
rs528015055 Chr.22: 37675799-37675799 on Build GRCh38
rs948744108 Chr.22: 37675696-37675696 on Build GRCh38
rs964774095 Chr.22: 37675854-37675854 on Build GRCh38
rs1028151730 Chr.22: 37675877-37675877 on Build GRCh38
rs552349358 Chr.22: 37675816-37675816 on Build GRCh38
rs112814081 Chr.22: 37675713-37675713 on Build GRCh38
rs12403 Chr.22: 37675700-37675700 on Build GRCh38
rs867863011 Chr.22: 37675734-37675734 on Build GRCh38
rs567137153 Chr.22: 37675759-37675759 on Build GRCh38
rs1004499643 Chr.22: 37675896-37675896 on Build GRCh38
rs1046541490 Chr.22: 37675726-37675726 on Build GRCh38
rs374036530 Chr.22: 37675725-37675726 on Build GRCh38
rs568526557 Chr.22: 37675794-37675794 on Build GRCh38
rs996627575 Chr.22: 37675869-37675869 on Build GRCh38
rs1047712559 Chr.22: 37675780-37675780 on Build GRCh38
rs974062985 Chr.22: 37675724-37675724 on Build GRCh38
rs945132996 Chr.22: 37675723-37675723 on Build GRCh38
rs995523869 Chr.22: 37675766-37675766 on Build GRCh38
rs113281514 Chr.22: 37675710-37675710 on Build GRCh38
rs770125794 Chr.22: 37675725-37675725 on Build GRCh38

View Full Product Details