Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25274441-25274686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYGB
Entrez Gene ID
5834
Gene Name
phosphorylase, glycogen; brain
Gene Aliases
GPBB
UniGene
Hs.368157
Gene Chromosome Location
Chr.20: 25248070-25298012 on Build GRCh38
Transcripts
NM_002862.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857532 c.613G>A p.G205R Chr.20: 25274676-25274676 on Build GRCh38
6760765 c.563A>G p.N188S Chr.20: 25274626-25274626 on Build GRCh38
4976906 c.559G>A p.G187S Chr.20: 25274622-25274622 on Build GRCh38
6760770 c.617G>A p.R206H Chr.20: 25274680-25274680 on Build GRCh38
6008543 c.571G>A p.E191K Chr.20: 25274634-25274634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986862331 Chr.20: 25274521-25274521 on Build GRCh38
rs549596880 Chr.20: 25274582-25274582 on Build GRCh38
rs377289028 Chr.20: 25274661-25274661 on Build GRCh38
rs200366571 Chr.20: 25274663-25274663 on Build GRCh38
rs199584158 Chr.20: 25274542-25274542 on Build GRCh38
rs73101785 Chr.20: 25274498-25274498 on Build GRCh38
rs760061144 Chr.20: 25274543-25274543 on Build GRCh38
rs756097079 Chr.20: 25274660-25274660 on Build GRCh38
rs910632819 Chr.20: 25274465-25274465 on Build GRCh38
rs200539270 Chr.20: 25274596-25274596 on Build GRCh38
rs756152089 Chr.20: 25274597-25274597 on Build GRCh38
rs942059525 Chr.20: 25274470-25274470 on Build GRCh38
rs1037876142 Chr.20: 25274514-25274514 on Build GRCh38
rs955385955 Chr.20: 25274518-25274518 on Build GRCh38
rs560745480 Chr.20: 25274511-25274511 on Build GRCh38
rs767380552 Chr.20: 25274655-25274655 on Build GRCh38
rs775415698 Chr.20: 25274628-25274628 on Build GRCh38
rs201326100 Chr.20: 25274616-25274616 on Build GRCh38
rs200865925 Chr.20: 25274622-25274622 on Build GRCh38
rs765081829 Chr.20: 25274568-25274568 on Build GRCh38
rs911347659 Chr.20: 25274538-25274538 on Build GRCh38
rs202169415 Chr.20: 25274620-25274620 on Build GRCh38
rs765785979 Chr.20: 25274626-25274626 on Build GRCh38
rs764081091 Chr.20: 25274639-25274639 on Build GRCh38
rs201847086 Chr.20: 25274557-25274557 on Build GRCh38
rs530521988 Chr.20: 25274488-25274488 on Build GRCh38
rs200000139 Chr.20: 25274604-25274604 on Build GRCh38
rs750281024 Chr.20: 25274656-25274656 on Build GRCh38
rs550363057 Chr.20: 25274515-25274515 on Build GRCh38
rs776621382 Chr.20: 25274552-25274552 on Build GRCh38
rs55654244 Chr.20: 25274569-25274569 on Build GRCh38
rs6037077 Chr.20: 25274647-25274647 on Build GRCh38
rs763157593 Chr.20: 25274631-25274631 on Build GRCh38
rs369278819 Chr.20: 25274539-25274539 on Build GRCh38
rs113869731 Chr.20: 25274553-25274553 on Build GRCh38
rs199968470 Chr.20: 25274621-25274621 on Build GRCh38
rs967284132 Chr.20: 25274507-25274507 on Build GRCh38
rs369733817 Chr.20: 25274558-25274558 on Build GRCh38
rs1050540249 Chr.20: 25274461-25274461 on Build GRCh38
rs1011443407 Chr.20: 25274489-25274489 on Build GRCh38
rs35481467 Chr.20: 25274603-25274603 on Build GRCh38
rs367624177 Chr.20: 25274662-25274662 on Build GRCh38
rs747769681 Chr.20: 25274617-25274617 on Build GRCh38
rs757574513 Chr.20: 25274644-25274644 on Build GRCh38
rs770679776 Chr.20: 25274482-25274482 on Build GRCh38
rs771236699 Chr.20: 25274618-25274618 on Build GRCh38
rs143100005 Chr.20: 25274642-25274642 on Build GRCh38

View Full Product Details