Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108162074-108162311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAT
Entrez Gene ID
4863
Gene Name
nuclear protein, coactivator of histone transcription
Gene Aliases
E14, E14/NPAT, p220
UniGene
Hs.171061
Gene Chromosome Location
Chr.11: 108155280-108222638 on Build GRCh38
Transcripts
NM_001321307.1 NM_002519.2 XM_011542854.2 XM_017017802.1 XM_017017803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
922680 c.3038C>T p.S1013L Chr.11: 108162153-108162153 on Build GRCh38
7173913 c.3050C>T p.S1017L Chr.11: 108162141-108162141 on Build GRCh38
5676407 c.3043G>T p.G1015C Chr.11: 108162148-108162148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990350166 Chr.11: 108162252-108162252 on Build GRCh38
rs778487123 Chr.11: 108162125-108162125 on Build GRCh38
rs776753811 Chr.11: 108162152-108162152 on Build GRCh38
rs940984590 Chr.11: 108162195-108162195 on Build GRCh38
rs978211913 Chr.11: 108162225-108162225 on Build GRCh38
rs750252765 Chr.11: 108162227-108162228 on Build GRCh38
rs899187192 Chr.11: 108162218-108162218 on Build GRCh38
rs773529061 Chr.11: 108162200-108162200 on Build GRCh38
rs775724349 Chr.11: 108162109-108162109 on Build GRCh38
rs762836090 Chr.11: 108162231-108162233 on Build GRCh38
rs745548434 Chr.11: 108162126-108162126 on Build GRCh38
rs924014475 Chr.11: 108162237-108162237 on Build GRCh38
rs761681319 Chr.11: 108162161-108162161 on Build GRCh38
rs750137921 Chr.11: 108162278-108162278 on Build GRCh38
rs373374919 Chr.11: 108162216-108162216 on Build GRCh38
rs1045101210 Chr.11: 108162246-108162246 on Build GRCh38
rs912112823 Chr.11: 108162190-108162190 on Build GRCh38
rs771414825 Chr.11: 108162127-108162127 on Build GRCh38
rs765149476 Chr.11: 108162179-108162179 on Build GRCh38
rs764527937 Chr.11: 108162236-108162236 on Build GRCh38
rs569919259 Chr.11: 108162145-108162145 on Build GRCh38
rs775075122 Chr.11: 108162144-108162144 on Build GRCh38
rs199936135 Chr.11: 108162148-108162148 on Build GRCh38
rs1025011198 Chr.11: 108162157-108162159 on Build GRCh38
rs905307492 Chr.11: 108162265-108162265 on Build GRCh38

View Full Product Details