Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 17073997-17074268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REPS2
Entrez Gene ID
9185
Gene Name
RALBP1 associated Eps domain containing 2
Gene Aliases
POB1
UniGene
Hs.186810
Gene Chromosome Location
Chr.X: 16946691-17194274 on Build GRCh38
Transcripts
NM_001080975.1 NM_004726.2 XM_005274625.3 XM_005274626.3 XM_011545603.2 XM_011545604.2 XM_011545605.2 XM_011545606.2 XM_011545607.2 XM_011545609.2 XM_017029955.1 XM_017029956.1 XM_017029957.1 XM_017029958.1

SNP Detail

SNP ID Chromosome Location
rs762159483 Chr.X: 17074081-17074081 on Build GRCh38
rs761743096 Chr.X: 17074200-17074200 on Build GRCh38
rs766904624 Chr.X: 17074101-17074101 on Build GRCh38
rs916698999 Chr.X: 17074223-17074223 on Build GRCh38
rs776426876 Chr.X: 17074195-17074195 on Build GRCh38
rs1049180552 Chr.X: 17074061-17074061 on Build GRCh38
rs775450562 Chr.X: 17074203-17074203 on Build GRCh38
rs776927958 Chr.X: 17074080-17074080 on Build GRCh38
rs748795997 Chr.X: 17074189-17074189 on Build GRCh38
rs181955231 Chr.X: 17074169-17074169 on Build GRCh38
rs189145906 Chr.X: 17074082-17074082 on Build GRCh38
rs755307277 Chr.X: 17074132-17074132 on Build GRCh38
rs376410889 Chr.X: 17074196-17074196 on Build GRCh38
rs921945224 Chr.X: 17074019-17074019 on Build GRCh38
rs750505958 Chr.X: 17074083-17074083 on Build GRCh38
rs373838317 Chr.X: 17074099-17074099 on Build GRCh38
rs1346246 Chr.X: 17074029-17074029 on Build GRCh38
rs771878060 Chr.X: 17074076-17074076 on Build GRCh38
rs762591782 Chr.X: 17074130-17074131 on Build GRCh38
rs368495237 Chr.X: 17074139-17074139 on Build GRCh38
rs745464800 Chr.X: 17074033-17074033 on Build GRCh38
rs747755239 Chr.X: 17074151-17074151 on Build GRCh38
rs755744458 Chr.X: 17074168-17074168 on Build GRCh38
rs752064144 Chr.X: 17074130-17074130 on Build GRCh38

View Full Product Details