Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23463812-23464069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIOK3
Entrez Gene ID
8780
Gene Name
RIO kinase 3
Gene Aliases
SUDD
UniGene
Hs.445511
Gene Chromosome Location
Chr.18: 23452823-23483140 on Build GRCh38
Transcripts
NM_003831.3 XM_011526242.1 XM_011526243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420902 c.238C>A p.L80M Chr.18: 23464025-23464025 on Build GRCh38
1750469 c.231C>T p.D77D Chr.18: 23464018-23464018 on Build GRCh38
3735535 c.232C>T p.L78F Chr.18: 23464019-23464019 on Build GRCh38
5914013 c.194C>T p.P65L Chr.18: 23463981-23463981 on Build GRCh38
473679 c.186T>C p.A62A Chr.18: 23463973-23463973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042321493 Chr.18: 23463888-23463888 on Build GRCh38
rs761333982 Chr.18: 23463973-23463973 on Build GRCh38
rs753005013 Chr.18: 23464025-23464025 on Build GRCh38
rs750212872 Chr.18: 23464037-23464037 on Build GRCh38
rs985657744 Chr.18: 23464018-23464018 on Build GRCh38
rs767993133 Chr.18: 23463952-23463952 on Build GRCh38
rs758599271 Chr.18: 23464031-23464031 on Build GRCh38
rs752539261 Chr.18: 23464008-23464018 on Build GRCh38
rs765191485 Chr.18: 23464022-23464022 on Build GRCh38
rs781024819 Chr.18: 23464033-23464033 on Build GRCh38
rs532897408 Chr.18: 23463994-23463994 on Build GRCh38
rs766473769 Chr.18: 23463938-23463938 on Build GRCh38
rs368949157 Chr.18: 23463937-23463937 on Build GRCh38
rs754244457 Chr.18: 23464006-23464006 on Build GRCh38
rs945752146 Chr.18: 23463886-23463886 on Build GRCh38
rs759703984 Chr.18: 23463970-23463970 on Build GRCh38
rs765792971 Chr.18: 23464011-23464011 on Build GRCh38
rs760860635 Chr.18: 23463835-23463835 on Build GRCh38
rs773688065 Chr.18: 23463971-23463971 on Build GRCh38
rs761329109 Chr.18: 23463967-23463967 on Build GRCh38
rs187456685 Chr.18: 23463929-23463929 on Build GRCh38
rs372420630 Chr.18: 23463957-23463957 on Build GRCh38
rs907457016 Chr.18: 23463889-23463889 on Build GRCh38
rs543987759 Chr.18: 23463858-23463858 on Build GRCh38
rs201943847 Chr.18: 23463942-23463942 on Build GRCh38

View Full Product Details